- Enables safe handling of dangerous materials or microorganisms
- Prevent moisture absorption while handling materials
- Provide personal protection from harmful substances
Gloved containment chambers offer you a safe, clean workspace that's ideal for enclosing balances or scales. HEPA and organic vapor filter safety valves protect you from particulates and harmful vapors. Four gas purging valves enable operation of weighing and transfer chambers. Top section is easily removable for equipment transfer.
What's Included:
one pair of size 9-1/2 chemical-resistant Hypalon® gloves; HEPA and organic vapor filters; a pressure relief valve; and a UL-listed, CSA-certified, 110 VAC electrical outlet strip (four outlets).
Additional Dimensions:
Transfer Chamber: 12" W x 11" I.D.
Glove Ports: 8¾" dia
Glove size: 9½ x 32 L