log in or register
Weller professional butane soldering pen kit
EW-97105-61
$95.00 USD/EACH
Usually ships in 10 days.
Qty
  • Soldering pen equivalent to a 25 to 75 watt soldering iron
  • ESD safe to protect sensitive components
  • Pen has an automatic cut-off switch when the cap is replaced
This lightweight soldering pen features professional grade quality in a compact design. Operate as a soldering iron or a heat tool. Gas catalyst system produces predictable and repeatable temperatures to ensure professional results. Convenient push-button switch offers quick and convenient ignition. Torch is powered by butane; order separately below.
What's Included:
Soldering tool, solder tip, flame tip, hot knife tip, hot air tip, hot air deflector, cap, tip stand, sponge and carrying case.
Product Type Self Igniting
Description Weller Professional Butane Solder Kit
Manufacturer number P2KC
Model 037103169372
Accessories
Qty/Price
Availability
at $47.50 USD/CASE
In stock.
at $18.00 USD/EACH
Usually ships in 11 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title