Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Sorry, this item is now discontinued.browse similar itemsor Contact us for more information.
West condenser, medium length, 190 mm, 19/22 joint
Kimax  |   Mfr# 282460-0200  |   Item# EW-34705-96
$65.00 USD/EACH
Discontinued
Qty
    Medium- length West condensers are ideal for refluxing and distillation assemblies. Design lessens condensate trapping.Full-length West condensers feature a stadard taper outer joint at he top and a standard taper inner drip joint at the bottom. High cooling efficiency is achieved with the increased flow rate of the cooling medium due to the design of the narrow annular space.
    Product Type Condensers, West
    Manufacturer number 282460-0200
    Brand Kimax
    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456