United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 10
of
32 >
Representative photo only
Wood Laboratory Table with Epoxy Top 48 x 24 x 36H
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-41530-18
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Furniture & Fume Hoods
>
Benches and Tables
>
Diversified Woodcrafts Laboratory Tables
Share
|
Wood Laboratory Table with Epoxy Top, 48 x 24 x 36H
EW-41530-18
$1,146.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Designed for the laboratory or classroom
Solid wood construction
Choose worksurface of black epoxy or chemguard laminate
For many a laboratory bench is one of the most important instruments a lab can have. Cleanliness, organization, and even results are all called into question when the laboratory bench isn’t respected as it should be. For comfort you could pick up a laboratory bench made from the standard solid wood construction with an epoxy resin / laminate work surface. A focus on clean perfection would warrant the stainless steel work table. Some of the stainless steel come factory installed rear back splashes. The standard economical versions, NSF approved ones and even ones that are approved for use in clean rooms. For help in choosing the right laboratory bench, speak with a Cole-Parmer Technical Expert.
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title