log in or register
YSI Model 550A Dissolved Oxygen Meter, 12-ft (3.7-m) probe cables
EW-53010-30
$875.00 USD/EACH
  • Polyethylene membrane reduces stirring requirements and improves response time
  • Innovative field-replaceable dissolved oxygen (DO) electrode modules save time and money
  • Waterproof, impactresistant case—IP67 rated
These DO meters feature a built-in calibration chamber that provides a 100% water-saturated environment—perfect for calibrating, transporting, and storing probe. Provide simultaneous readings of oxygen and temperature. Measurements are fast and easy with one-handed operation, large backlit display, automatic push-button calibration, and a weighted, quick-sinking probe. Low-battery indicator alerts you to change batteries. Salinity, temperature and altitude compensations eliminate the need for correlation chart.
Product Type Dissolved Oxygen Meter
Meter style Portable
Dimensions (" W) 4-3/8
Dimensions (" H) 9
Dimensions (" D) 3-1/2
Range (Dissolved oxygen) 0 to 50 mg/L
Range (Temperature °C) 5 to 50
Range (Temperature °F) 23 to 113
Resolution (Temperature) 0.1
Resolution (Dissolved Oxygen) 0.01 mg/L
Temperature compensation Automatic
Display LCD
Waterproof Yes
CE Compliance Yes
Accessories
Qty/Price
Availability
at $12.55 USD/PACKAGE of 4
In stock.
at $195.00 USD/EACH
Usually ships in 10 days.
at $28.00 USD/EACH
Usually ships in 14 days.
Ultra-C Mount Clamp
EW-99852-68
at $26.50 USD/EACH
Usually ships in 14 days.
at $56.00 USD/EACH
In stock.
Calibrations
Qty/Price
Availability
at $115.00 USD/EACH
Usually ships in 16 days.
Technical Resources
  • Instruction Manual
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title