log in or register
Yamato Vertical-Loading Portable Sterilizer; capacity, 1.2 cu ft; chamber, 17 1/2
EW-10771-07
$7,585.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
  • Vertical loading maximizes usable chamber space
  • Casters and flexible plumbing make sterilizers completely mobile
  • Water-resistant keypad protects against steam and condensation
  • Front door water level site glass shows you when the water tank needs to be emptied
These compact portable sterilizers are ideal for laboratories that share equipment. Single-cycle models sterilize while dual-cycle models feature both sterilizing and drying cycles. Designed with rolling casters for excellent mobility and easy storage—just push it into a storage room when not in use to make room in your lab. Top mounted stainless steel door also saves space in your lab and provides easy loading.
The sterilizers feature fully automated operation. Simply place your load in the pressure chamber and enter all parameters with the easy-to-use keypad. The LED display shows set and actual temperature, as well as set time and time remaining.
Self-diagnostic circuitry monitors the temperature sensor, low-water sensor, and heater; alerts you with an audible and a visual alarm if a malfunction occurs. Additional safety features include a circuit breaker, safety pressure release valve, and a water level site glass.
What's Included:
Two standard, stackable baskets and a 5-ft cord with three-prong plug (models 10772-07 and -15 do not include a plug). Solid baskets with side vents and baskets with adjustable shelves are also available; order separately.
Product Type Sterilizers
Capacity (cu ft) 1.2
Reservoir capacity (Liters) 3
Chamber Capacity (Liters) 34
Chamber size (" H) 17-1/2
Chamber size (" Dia) 11-13/16
Chamber size (cm) 44.5 H x 30 dia
Max temperature (º C) 128
Min temperature (º C) 105
Min temperature (º F) 221
Max temperature (º F) 262
Dimensions (" W) 17-5/16
Dimensions (" H) 33-13/16
Dimensions (" D) 20-15/16
Dimensions (cm) 44 W x 85.9 H x 53.2 D
Timer range 1 mins - 99 hrs & 59 mins; 100 - 999 hrs
Power (VAC) 220
Power (Hz) 50/60
Power (Amps) 11
Chamber construction 304 Stainless Steel
Display LED
Max psi 29
Manufacturer number SM-310
Brand Yamato
Accessories
Qty/Price
Availability
at $81.00 USD/BOX of 100
In stock.
at $100.00 USD/PACKAGE
In stock.
Plastic tie, 4"L
EW-06831-15
at $15.75 USD/PACKAGE of 1000
In stock.
Autoclavable Tape
EW-08277-62
at $8.75 USD/ROLL
In stock.
at $29.00 USD/BOX of 100
In stock.
at $29.00 USD/BOX of 100
Usually ships in 6 days.
at $335.00 USD/EACH
Usually ships in 15 days.
at $585.00 USD/EACH
Usually ships in 15 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.
    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title