Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

inoLab 7310 advanced pH/mV/T benchtop meter with printer

WTW  |   Item# WU-58890-64

$1,307.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Available for order
or call to order
Qty

  • Providing accurate measurements, compliant documentation, and secure traceability
  • Auto-recognition of USA/NIST and DIN buffers
  • AutoRead function ensures repeatable results
  • Easy to calibrate and maintain with calibration timer
  • Power with universal AC adapter or AA batteries
The inoLab family of benchtop meters offer an intuitive user interface, large informative display, and keypad with tactile feedback to provide an exceptional user experience for error-free operation.

inoLab pH 7310 Advanced Meter allows accurate measurements with compliant documentation to support GLP (Good Laboratory Practice) and AQA (Analytical Quality Assurance) laboratory requirements. Capture 500 manual or 5000 automatic data sets with time/date stamp, sample ID, and calibration data. USB interface provides an easy connection to a PC, or choose the model with integrated printer for immediate results. All data is transfered in a CSV format directly into Excel®.


What's Included:
Electrode stand, universal power adapter, and four AA batteries. Models inoLab pH 7310 and inoLab Multi 9310 also include data logging software and cable. Purchase electrodes separately.
Product Type pH Meter
Meter style Benchtop
Range (pH) -2.000 to 20.000
Range (mV) ±1200
Range (Temperature °C) -5 to 105
Range (Temperature °F) 23 to 221
Resolution (pH) 0.1/0.01/0.001
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Accuracy (pH) ±0.005 pH
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ± 0.3 mV, ± 1 digit
Accuracy (Dissolved Oxygen) ±0.2mg/L from 0 to 20 mg/L; ±0.5 mg/L from 50 to 50 mg/L
Temperature compensation Automatic or manual
Display LCD
Interface USB
Calibration Up to 5 points
Waterproof Yes
Qty/ea 1
Buffer recognition USA, NIST, DIN
Power AC adapter or four AA batteries(optional)
Datalogging (points) 5000
CE Compliance Yes
Brand WTW
Dimensions (" H) 7.5
Dimensions (" D) 3.125
Dimensions (" L) 9.5
Model PH 7310 w/printer

Accessories
Qty/Price
Availability

at $9.25 USD/PACKAGE of 4

In stock.

at $20.00 USD/EACH

Usually ships in 6 days.

at $275.00 USD/EACH

Usually ships in 9 days.

at $312.00 USD/EACH

Usually ships in 10 days.
Related products
Qty/Price
Availability

at $17.75 USD/PACKAGE
In stock.
Calibrations
Qty/Price
Availability

at $126.00 USD/EACH
Usually ships in 6 days.




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews