Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

pH Module only for iPhone, iPad, or iPod

Thermo Scientific Orion  |   Item# WW-05760-12

$205.00 USD/EACH
$194.75 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 8 days.
Qty

  • Convert your iPhone into a pH meter
  • Powered by the iPhone; no charging required
  • Data transfer by email or with USB connection
  • Calibrate meter with 2, 3, 4...up to 99 points
  • Automatic and manual temperature compensation; °C or °F
Simply plug the PH-1 module into your iPhone, iPad, or iPod to measure and record values, date and time, and GPS location. Display shows pH measurement, mV reading, ambient temperature, and ATC temperature. Take pH and ORP measurements utilizing any electrode with a BNC connector.

Follow easy on screen prompts to install free app, connect an electrode, select temperature compensation and calibrate.Purchase pH electrode separately.
What's Included:
iPhone, iPad, iPod NOT INCLUDED.
Product Type pH Module
Meter style Benchtop
Range (pH) 0.00 to 14.00
Range (mV) ± 1250
Range (Temperature °C) 0 to 100
Range (Temperature °F) 32 to 212
Resolution (pH) 0.1/0.01
Resolution (Temperature) 0.1
Resolution (mV) 1/10
Accuracy (pH) ±0.01 pH
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±1mV ±1 digit
Temperature compensation Automatic
Calibration Up to 5 points
Connections BNC connector
Qty/ea 1
Buffer recognition USA, NIST
Power Connect to i device
CE Compliance Yes
Brand Thermo Scientific Orion
Dimensions (" W) .5
Dimensions (" H) 2"
Dimensions (" L) 3 1/4"
Model MODULE

Accessories
Qty/Price
Availability

at $30.25 $19.99 USD/PACKAGE of 3

In stock.

at $141.00 USD/EACH

In stock.

at $102.00 $91.80 USD/EACH

In stock.
Related products
Qty/Price
Availability

at $17.75 USD/PACKAGE
In stock.




Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews