Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
pHydrion Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package
Phydrion  |   Mfr# P-12M  |   Item# EW-59200-32
$39.25 USD/PACKAGE
Quantity Discounts
Qty: 10 25 40+
Price: $35.33 $33.36 $31.40
  • Immediately gives the surface reaction of any solid to the nearest pH value
  • Retractable, nonrefillable, mechanical pencils are more convenient than paper-wrap pencils
  • The widest range available — reading from pH 0 to 13 — with 14 different color matches
The handy, easily portable form is perfect for pH spot checks, requiring as little as 0.001 mL of buffered solution. Use pencil on virtually any surface—even irregular or uneven ones. It is effective on porous material and impervious solids.
To use, first wet the surface or area with distilled water and stroke several times with the pH pencil. After 15 seconds, compare the resulting color against the color chart. Pack includes three mechanical pencils and two color charts.
Product Type Test Strip
Parameters pH
Description pHydrion Insta-Chek Mechanical pH Pencil, Pack of 3
Range 0-13 pH
Graduations 1
Model P-12M
Qty/pk 3
Manufacturer number P-12M
Brand Phydrion
Related products
Qty/Price
Availability
at $17.75 USD/PACKAGE
In stock.
Calibrations
Qty/Price
Availability
at $126.00 USD/EACH
Usually ships in 6 days.
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews
    REVIEW SNAPSHOT®
    by PowerReviews
    pHydrion Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package
    4.3
    (based on 3 reviews)
    100%
    of respondents would recommend this to a friend.
    Reviewed by 3 customers
    Sort by
    Displaying reviews 1-3
    Back to top
    5.0
    Good Product
    By ChantalD
    from Gwynn Oak, MD
    About Me Casual Do-It-Yourselfer
    Verified Buyer
    Pros
    • Easy To Use
    Cons
      Best Uses
      • Bathroom
      • Test products
      Comments about pHydrion Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package:
      The pencil lasts a long time and I'm sure I'll get plenty of use out of it. Great product. Just what I needed for my business.
      4.0
      Fine
      By Lynn
      from Baltimore MD
      About Me Professional
      Verified Buyer
      Pros
      • Attractive Design
      • Durable
      • Easy To Use
      Cons
      • More length to last longe
      Best Uses
      • Arbonne presentations
      • Office
      Comments about pHydrion Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package:
      This is the second time purchasing. I share it with other team members.
      (1 of 1 customers found this review helpful)
      4.0
      pHydrion Insta-Chek Mechanical pH Pencil
      By GP-CA
      from San Clemente, CA
      Verified Buyer
      Pros
        Cons
          Best Uses
          • Printing pH on slab
          • Water before or after
          Comments about pHydrion Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package:
          Better product now. Might be nice as a full chulk size.
          Displaying reviews 1-3
          Back to top
          Customers Who Bought This Item Also Bought