log in or register
pHydrion plastic pH indicator strips, 0.0 to 14.0, flip top vial packaging
EW-59200-40
$16.20 USD/PACKAGE
  • Three times the matching area compared to other plastic strips
Immediate pH testing is available in brilliant, long lasting colors. These direct-reading pH papers give a simple, single color match for all pH values in distinct, bright color separations. Handy flip top vials contain 100 dip sticks and feature a handy matching color chart.
Product Type Test strips
Parameters pH
Quantity 100
Range 0 - 14
Graduations 1
Accessories
Qty/Price
Availability
at $10.25 USD/EACH
In stock.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title