Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
1-800-323-4340
Call for pump building assistance
Or
Save time and money- let us custom cut your tubing
Masterflex C/L Series Tubing Pumps
Masterflex C/L Series Peristaltic Pumps
C/L® series peristaltic pumps combine low flow and compact size in one pump that sits on your benchtop or mounts in a rack. Ideal for analyzer applications.
C/L® Series Pump Components
    A single channel and
    quick-and-simple loading
    Two synchronous pumping channels
    for added versatility
    Available in sizes and formulations
    to fit your applications
  Masterflex Series
    High accuracy
    Flow Range:
    0.0006 to 3400 mL/min
    Powerful systems
    Flow Range:
    0.001 to 26 LPM
    Highest flow rates
    Flow Range:
    0.3 to 42 LPM