Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
1-800-323-4340
Call for pump building assistance
Or
Save time and money- let us custom cut your tubing
Masterflex peristaltic pump components - full systems
Masterflex Peristaltic Pump Tubing Systems
Enjoy the power and accuracy of a Masterflex peristaltic pump drive right out of the box, ready to go.
Complete Pump Systems
    Compact and low flow with single and dual channels; variable speeds.
    High accuracy for precise flow control. Ideal for laboratory, field or process applications
    Powerful motors and better protection from hazards. Ideal for process and industrial
    High flow rate, durable motors. Ideal for transfer of viscous or shear-sensitive materials
  Masterflex Series
    Compact, low flow
    Flow Range:
    0.002 to 43 mL/min
    High accuracy
    Flow Range:
    0.0006 to 3400 mL/min
    Powerful systems
    Flow Range:
    0.001 to 26 LPM
    Highest flow rates
    Flow Range:
    0.3 to 42 LPM