Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
1-800-323-4340
Call for pump building assistance
Or
Save time and money- let us custom cut your tubing
Masterflex I/P Series Peristaltic Tubing Pumps
Masterflex I/P Series Peristaltic Pumps
I/P® series peristaltic pumps have powerful motors and better protection from industry hazards. Ideal for manufacturing, process and industrial applications.
I/P® Series Pump Components
    Built for durability in process and industrial applications
    Custom manufactured to
    ensure accurate and repeatable performance
    Durable with remote I/O option. Easily integrates into process and industrial applications
    Convenient one-number systems are complete and ready to use
  Masterflex Series
    Compact, low flow
    Flow Range:
    0.002 to 43 mL/min
    High accuracy
    Flow Range:
    0.0006 to 3400 mL/min
    Highest flow rates
    Flow Range:
    0.3 to 42 LPM