Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
1-800-323-4340
Call for pump building assistance
Or
Save time and money- let us custom cut your tubing
Masterflex pump components - drives
Masterflex Peristaltic Pump Drives
Precision engineered Masterflex® peristaltic pump drives offer wide flow ranges and control options for applications from lab and field to process and industrial.
Pump Drives
    Fixed or variable speed control; console or modular design. High-accuracy digital drives offer precise transfer and dispensing
    Rugged designs and options for remote I/O control; easily integrate into process and industrial applications
  Masterflex Series
    Compact, low flow
    Flow Range:
    0.002 to 43 mL/min
    High accuracy
    Flow Range:
    0.0006 to 3400 mL/min
    Powerful systems
    Flow Range:
    0.001 to 26 LPM
    Highest flow rates
    Flow Range:
    0.3 to 42 LPM