Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
1-800-323-4340
Call for pump building assistance
Or
Save time and money- let us custom cut your tubing
Masterflex pump components - pump heads
Masterflex Peristaltic Pumps Heads
Engineered for long life, Masterflex® peristaltic pump heads provide superior accuracy and repeatability and ensure optimal performance when used with Masterflex® pump tubing.
Pump Heads
    Nine style options ensure that your
    application needs are met
    Durable for process
    and industrial applications
    Convert your existing motor into a high-capacity Masterflex B/T pump
  Masterflex Series
    Compact, low flow
    Flow Range:
    0.002 to 43 mL/min
    High accuracy
    Flow Range:
    0.0006 to 3400 mL/min
    Powerful systems
    Flow Range:
    0.001 to 26 LPM
    Highest flow rates
    Flow Range:
    0.3 to 42 LPM