Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
1-800-323-4340
Call for pump building assistance
Or
Save time and money- let us custom cut your tubing
Masterflex peristaltic pump components -  tubing
Masterflex Peristaltic Pump Tubing
Engineered for long life, Masterflex® peristaltic pump tubing provides superior accuracy and repeatability and ensures optimal Masterflex® pump performance.
Pump Tubing
    Precision-extruded microbore tubing delivers repeatable performance in C/L pumps
    Extremely close tolerances to match L/S pump heads, ensuring accurate, repeatable flow and long life
    Extremely close tolerances to match I/P pump heads, ensuring accurate, repeatable flow and long life
    B/T PerfectPosition™ tubing ensures proper loading and repeatable performance
  Masterflex Series
    Compact, low flow
    Flow Range:
    0.002 to 43 mL/min
    High accuracy
    Flow Range:
    0.0006 to 3400 mL/min
    Powerful systems
    Flow Range:
    0.001 to 26 LPM
    Highest flow rates
    Flow Range:
    0.3 to 42 LPM