United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Balances & Scales
Analytical Balances
Balance and Scale Accessories
Calibration Masses
Mechanical Balances and Scales
Microbalances
Moisture Balances
Pipette Accuracy Testers
Portable Balances
Scales
Toploading Balances
Baths & Circulators
Baths & Circulators
Bath & Circulator Accessories
Circulating Water Baths
Heated Water Baths
Laboratory & Utility Water Baths
Orbital Motion Heated Bath
Recirculators, Chillers, Immersion Chillers and Coolers
Shaking Water Baths
Tanks for Baths and Circulators
Calibration
Calibration
Calibration Equipment
Calibration Masses
Calibration Services
Instrument Calibration
Chemicals, Reagents, & Lubricants
Chemicals, Reagents, & Lubricants
Agar and Agarose
Buffers, Storage Solutions, and Standards
Chemicals
Cleaning Chemicals
Lubricants
Reagents
Specialty Chemicals
Water
Cleaning & Janitorial Supplies
Cleaning & Janitorial Supplies
Biohazard Bags and Biosafety Disposal
Cleaner/Degreasers and Contact Cleaners
Cleaning Brushes
Cleaning Solutions, Detergents, & Skincare
Mop Handles
Mops
Pails
Spray Bottles
Stainless Steel Scrub Pads
Cooling Equipment
Cooling Equipment
Cryogenics, Cryogenic Dewars, and Cryogenic Storage Products
Freezers - Chest/Cabinet
Freezers - Upright freezers
Ice Machines
Recirculators, Chillers, Immersion Chillers and Coolers
Refrigerator and Freezer Combination
Refrigerators
Critical Environment
Critical Environment
Air Sampling
Airflow Measurement
Cleaners
Cleanroom and Static Control Gloves
Cleanroom Furniture
Critical Environment Apparel
Mats
Mops
Sanitary Scrub Brushes
Stationary
Vacuums
Wipes and Swabs
Flow, Level & Valves
Flow, Level & Valves
Flowmeters
Level Control
Valves
Fluid Handling Equipment
Fluid Handling Equipment
Centrifugal Separation
Masterflex
®
Peristaltic Pumps, Tubing, & Accessories
Microfluidics
Mixers, Blenders, and Stirrers
Pumps
Pumps - Centrifugal Pumps
Pumps - Drum and Hand Pumps
Pumps - Fillers
Pumps - Gear Pumps
Pumps - Ismatec
®
Pumps - Manostat Peristaltic Pumps
Pumps - Metering Pumps
Pumps - Positive Displacement Pumps
Pumps - Systems
Pumps - Vacuum Pumps
Furniture & Fume Hoods
Furniture & Fume Hoods
Benches and Tables
Carts, Mobile Cabinets, and Mobile Workstations
Chairs and Stools
Fume Extractors
Fume Hoods
Glove Boxes
Junction Boxes
Mobile Cabinet
Shelving
Wall Mount Enclosures, Cabinets, & Racks
Workstations
Heating Equipment
Heating Equipment
Block Heaters
Burners
Drum Heaters
Environmental Chambers
Furnaces
Heat Guns
Heat Sealer
Heating Mantles
Heating Tapes, Cords, Blankets
Heating Torches
Hot Plates
Immersion Heaters
Incubators
Ovens
Specialty Heaters
Sterilizers
Lab Supplies & Consumables
Lab Supplies & Consumables
Apparel
Bags
Beakers
Books & Reference Materials
Bottles
Bowls
Brushes
Cable Ties
Calculators
Carboys
Clamps and Supports
Cleaning Solutions, Detergents, & Skincare
Containers
Crucibles
Dishes
Film and Foil
Filtration
Flasks
Funnels
Glassware
Graduated Cylinders
Ice Buckets and Ice Pans
Label Printers
Labels
Laboratory Organizers
Labware
Ladles and Dippers
Mattings and Liners
Mortars & Pestles
Notebooks
Pails
Pens and Markers
Racks
Sample Containers
Sanitary and Sterile Products
Scoops and Spoons
Shippers
Spatulas & Stir Rods
Stainless Steelware
Stir Bars
Stoppers
Tapes
Test Tubes
Timers
Tongs and Tweezers
Trays
Vials
Laboratory Equipment
Laboratory Equipment
Balances & Scales
Baths & Circulators
Centrifuges
Clamps and Supports
Desiccators
Dispensers
Electrophoresis Products
Evaporators
Extraction
Filtration
Flame Photometers
Fluid Bed Dryers
Freeze Dryers
Glassware Washers
Heating Equipment
Humidifiers/Dehumidifiers
Laser Products
Lighting and Illumination
Microscopes & Magnification
Mixers, Blenders, and Stirrers
Pipettes and Pipettors
Refractometers
Samplers
Shakers
Sieves
Stainless Steelware
Timers
Ultrasonic Cleaners
Ultrasonic Processors
Ultraviolet Equipment
Viscometers
Water Purification Systems and Accessories
Life Science
Life Science
Animal Research Products
Cell Biology
Chromatography Products
Dialysis
Electroporation
Fermentation
Forensics
Genomics
High Throughput Screening
Histology
Immunochemistry
Immunology
Microbiology
Microfluidics for Life Science Applications
Molecular Biology
PCR (Polymerase Chain Reaction)
Proteomics
Spectroscopy and Spectrophotometers
Stem Cell Research
Synthesis Systems
Tissue and Cell Culture
Western Blotting
Liquid Handling
Liquid Handling
Burettes (Burets)
Dispensers
Pipettes and Pipettors
Piping
Syringes
Titrators
Masterflex
®
Peristaltic Pumps, Tubing, & Accessories
Masterflex
®
Peristaltic Pumps, Tubing, & Accessories
InnoCal
®
Pump Calibration and Repair Services
Masterflex Accessories
Masterflex Diss-Washer
Mobile Wash Station
Masterflex
®
B/T
®
Tubing Pumps
Masterflex
®
C/L
®
Tubing Pumps
Masterflex
®
fittings
Masterflex
®
I/P
®
Tubing Pumps
Masterflex
®
L/S
®
Tubing Pumps
Masterflex
®
Pump Head Adapters
Masterflex
®
Replacement Parts
Masterflex
®
Self-Contained Composite Sampler
Masterflex
®
Tubing
Masterflex
®
Tubing Formulations
Material Storage and Handling
Material Storage and Handling
Carts, Mobile Cabinets, and Mobile Workstations
Document and Waste Containers
Enpac Cylinder Dolly
Hand & Platform Trucks
Instrument Cases
Pelican Watertight Instrument Cases
Plastic Storage Bins & Containers
Stools & Ladders
Storage and Utility Cabinets
Worktables and Trucks
Material Testing / Physical Testing
Material Testing / Physical Testing
Color & Gloss Meters
Density/Specific Gravity
Force & Torque
Melting Point
Physical Property Testing
Polarimeters
Relative Humidity
Surface Testing
Tensiometers
Microscopes & Magnification
Microscopes & Magnification
Cameras and Video Microscopy Equipment
Compound Microscopes
Fiberscopes and Borescopes
Illuminated Magnifiers
Magnifiers
Microscope Accessories
Microscope/Telescopes (Dual-Purpose)
Shop Microscopes
Specialty Microscopes
Stereo and Stereozoom Microscopes
Wide-Field Microscopes
Mixers, Blenders, and Stirrers
Mixers, Blenders, and Stirrers
Blenders and Paddle Blenders
Dispersion Mixers and Accessories
Handheld Mixers
Handlebar Pail and Drum Air Mixers
Homogenizers
IKA Magic LAB Mixing System
Industrial Mixers
Laboratory Mixers
Lightnin
®
Mixers and Accessories
Mills
Pail Mixer with Rim Clamp
Powder Mixers
Sanitary Mixers
Static (In-line) Mixers
Stir Bars
Stirrers
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Accessories for pH Meters and Ion Meters
Buffers, Storage Solutions, and Standards
Chemical Oxygen Demand Testing
Chloride Analyzers
Colorimeters
Colorimetry Reagents
Conductivity/Resistivity/TDS Meters and Testers
Dissolved Oxygen (DO) Probes and Sensors
Dissolved Oxygen Benchtop Meters
Dissolved Oxygen Handheld Meters
Dissolved Oxygen Pocket Testers
Dissolved Oxygen Test Kits
Dye Tracers
Electrodes
Multiparameter Meters
pH Meters, Ion Meters and ISFET Meters
pH Test Strips, pH Indicators, and Water Quality Testing Kits
Process Equipment, Process Instruments & Process Controllers
Turbidity Meters and Turbidity Testing Products
Plastic Tubing
Plastic Tubing
Flexible Tubing
Heat-Shrink Tubing
Hose
Microbore Tubing
Rigid Tubing
Tubing Clamps, Tubing Cutters, and Accessories
Pressure & Vacuum Instruments
Pressure & Vacuum Instruments
Air Compressors
Calibrators
Fluke 700LTP-1 Hand Operated Low Pressure Pump
Fluke-700PTP-1 Pneumatic Test Pump
Gauge guards, Seals, and Accessories
Gauges
Manometers
Presses
Pressure & Vacuum Controllers
Pressure I/P Transducers
Pressure Regulators - Industrial
Pressure Regulators for Gases
Pressure Transmitters and Pressure Transducers
Sanitary Transducers, Gauges, and In-Line Seals
Switches
The Fluke 700HTP-1 Hand Operated Hydraulic Pressure Pump
The Fluke 700PMP Hand-Operated Pressure Pump
Vacuum Controllers, Gauges, and Systems
Water Pressure Loggers
Process Equipment & Instrumentation
Process Equipment & Instrumentation
Actuators
Automation Control
Electrical and Electronics
Flowmeters
Level Control
Panel Meters
Process Controllers
Pumps
Steam Generators
Steam Handling
Tachometers/Stroboscopes
Tanks
Valves
Vibration Measurement
Pump Systems
Pump Systems
Air Compressors
Motors
Pumps
Recorders & Data Acquisition
Recorders & Data Acquisition
Data Acquisition
Dataloggers
Pressure Dataloggers with LCD Display
Recorders
Safety Products
Safety Products
Air Sampling and Gas Analysis
Alarms
Arc Flash Products
Confined Space Gas Detectors
Electrical Safety
Ergonomics
Eyewash
Eyewear and Safety Glasses
First Aid
Floor Mats and Matting
Gloves
Hard Hats
Heat Stress Monitors
Light Meters
Lighting
Lockout/Tagout Product
Metal Detectors
Personal Protective Equipment (PPE)
Radiation Detectors
Radios
Respirators and Masks
Safety Apparel
Safety Light Curtains
Safety Shields
Safety Signs & Labels
Safety Software
Sound Meters
Spill Control
Storage and Handling
Shop by Industry
Shop by Industry
Biofuels
Biopharma
Chemical Property Testing
Environmental Testing
Food Quality & Testing
Health and Safety
Industrial Process
Laboratory Research
Life Science
Natural Resources and Mining
pH Measurement
Pharmaceutical Industry Products
Weather Testing Equipment
Single - Use Technology
Single - Use Technology
Check and Foot Valves with Hose Barb Connections
Infrared PTFE, PVDF and Stainless Steel Turbine Flow Sensors
Luer Stopcocks
Miniature Check Valves
Overlook Industries
®
OneShot
Single-Use Filler Nozzles
Tubing for Bioprocessing
Valves
Temperature, Humidity and Moisture Products
Temperature, Humidity and Moisture Products
Humidity Indicators (Dial)
Humidity Indicators (Digital)
Humidity Loggers and Recorders
Humidity Meters (Thermohygrometers Meters)
Humidity Pens (Thermohygrometer Pen-style)
Humidity Psychrometers / Mason Hygrometers
Humidity Switches / Panel Mount Controllers / Alarms/Monitors
Humidity Transmitters
Hydrometers
Moisture Balances
Moisture Meters
Moisture Water Activity Systems
Temperature Bimetal Thermometers
Temperature Chart Recorders
Temperature Controllers
Temperature Dataloggers
Temperature Digital Thermometers
Temperature Glass Thermometers
Temperature Infrared Thermal Imagers
Temperature Infrared Thermometers / Sensors
Temperature Labels and Crayons
Temperature Probes and Accessories
Temperature RTD Instruments and Probes
Temperature Thermistor Instruments and Probes
Temperature Thermocouple Instruments and Probes
Weather Testing Equipment
Tools
Tools
Adhesives and Epoxies
Calipers
Hand Tools
Inspection / Pick-up Tools
Measuring Tools
Micrometers
Precision Indicators
Silicone Spray
Soldering Tools
Stationary and Portable Vises
Tool Kits
Tubing, Fitting, & Hose
Tubing, Fitting, & Hose
Clamps for Tubing
Fittings for Tubing and Hose
Gaskets
Hose
Tubing
Valves
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Products
Alphabetically (A-Z)
Category Index
Industry Index
New Products
Best Sellers
Specialty Brands
Virtual Catalog (PDFs)
Special Offers
Clearance Items
Guaranteed in Stock
Customized Solutions
Special Order Items
Custom-blended Chemicals
Sterile Single-use
Assemblies - BioConnect
®
Temperature Probe
Configurator - Oakton
®
Pump Configurator - Masterflex
®
Custom Safety Signs
Item Number Ordering
Quick Order Form
My Shopping Lists
Free Catalogs
Shopping Assistance
Product Selection Guides
Sign up for Special Offers
Subscribe Today
Need Expert Advice?
1-800-323-4340
E-mail Us
Live Chat
Expert Assistance with
Product Selection, Application, Technical
Advice, and Troubleshooting
Special Ordering/Product Modifications
Customer Service
Call Center Hours: 7am - 6pm CST
(Technical Support Hours: 8am - 6pm CST)
Services
Calibration - InnoCal
Repairs
Leasing Program
Returns & Cancellations
Warranty Policy
Shipping Policy
Site Support
Contact Us
1-800-323-4340
E-mail Us
Live Chat
Tools
Chemical Compatibility Database
Safety Glove Chemical Compatibility Database
Masterflex
®
Tubing and Chemical Compatibility
Unit Converter
Conversion Factors and Tech Data
Technical Glossary
Guides
Product Selection Guides
Product Maintenance Guides
Product FAQs
Video
Technical Resource Library
Library Main
Instruction Manuals
MSDS
Articles and White Papers
Case Studies
Links to Regulatory Agencies
Masterflex
®
Application Solutions
Stay Informed of the Latest News...
Subscribe Today
Technical Experts are Ready to Help
1-800-323-4340
E-mail Us
Live Chat
My Account
My Account Profile
Login
Forgot Login ID or Password?
Create a Profile (Register)
My Shopping Lists
My Saved Addresses
Add a New Address
Communication Settings
Preferences and Subscriptions
Track your order, convert quotes,
and reprint invoices.
My Transactions
My Transaction History
Look Up My...
Order, Quote, or Invoice
Why Register with Us?
Express
Register
Technical Resources
You are filtering by:
Video
Clear all filters
Technical Resource Map
Looking for chemical compatibility ratings?
Use our searchable Chemical Compatibility Database
Need assistance?
Contact us
for
Expert Tech
Advice
Product
Selection
Custom
Solutions
Free Catalogs
Choose one for your specific needs.
Request your copy today.
Share
|
Home
>
Technical Resources
>
Video
Technical Resource Library
Sort by
Best Match
Name
Recent Post Date
Search
0
results (showing
1
-
0
)
Per Page
5
10
15
25
50
Close
Gear pump head replacement
View products by:
Job Title
|
Category
|
Brand
|
Part Number
|
Price
View product categories by:
Popularity
|
Favorites
|
Job Title
|
Price
|
Part Number
View popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Job Title
View reviews by:
Popularity
|
Product Title
|
Best Rated