United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths & Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cleaning & Janitorial Supplies
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Lab Supplies & Consumables
Laboratory Equipment
Life Science
Masterflex® Peristaltic Pumps
Masterflex® Pump Tubing
Material Storage & Handling
Material Testing & Physical Testing
Microscopes & Magnification
Mixers and Blenders
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity and Moisture Products
Tools
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Technical Resources
You are filtering by:
Video
Clear all filters
Industries:
Electrochemistry
(1)
Environmental
(2)
Food and Beverage
(2)
[+]
Show more
(4)
Petrochemical
(2)
Process & Control
(1)
Transportation
(2)
Water and Wastewater
(1)
[-]
Show less
Product Resources:
pH Meters
(1)
Technical Resource Map
Looking for chemical compatibility ratings?
Use our searchable Chemical Compatibility Database
Need assistance?
Contact us
for
Expert Tech
Advice
Product
Selection
Custom
Solutions
Free Catalogs
Choose one for your specific needs.
Request your copy today.
Share
|
Home
>
Technical Resources
>
Video
Technical Resource Library
Sort by
Best Match
Name
Recent Post Date
Search
Video
Measuring pH of soil
In this video we show you how to take a pH measurement of your soil samples. The items we used were the pH 11 meter (35614-22), temperature probe (35613-05), and electrode 35805-01.
Apr 17 2012 11:06AM
Video
Video - How to Calibrate the Oakton pH Testr
This helpful video will show you how to quickly calibrate your Oakton Instruments pH Testr.
Apr 4 2012 3:39PM
Video
How to Load tubing into Masterflex L/S standard pump heads
This videos shows how to load tubing into the Masterflex L/S standard pump heads.
Nov 11 2011 2:56PM
Video
How to load Masterflex 91BT pump head tubing
This video shows how to load 87BT pump head tubing into the Masterflex BT peristaltic pump.
Nov 11 2011 2:48PM
Video
How to load Masterflex 87BT pump head tubing
This video shows how to load 87BT pump head tubing into the Masterflex BT peristaltic pump.
Nov 11 2011 2:58PM
Video
Gear pump head replacement
This video shows how to perform gear pump head replacement.
Nov 11 2011 2:59PM
Video
DO Probe (filling solution, membrane replacement and calibration)
This video demonstrates general DO probe calibration and maintenance
Jun 2 2011 12:00AM
Video
Masterflex Digital Drive Calibration
This video shows how to calibrate a Masterflex peristaltic pump.
May 25 2011 12:00AM
Video
pH Electrode Maintenance
This video shows how to clean and spin your electrode and shorting your pH meter.
May 10 2011 12:00AM
Video
Serial Dilution -- How to perform a serial dilution
This video shows how to perform a serial dilution of a 1000ppm standard down to 1pm.
Mar 31 2011 12:00AM
Video
Measurement of fluoride concentration in tap water
This video shows how to calibrate your fluoride tester and measure fluoride concentration in tap water with our handheld fluoride pocket tester.
Mar 25 2011 12:00AM
Video
Variable Area Flowmeters
This video shows how to operate our variable area flowmeters
Mar 1 2011 12:00AM
Video
Oakton TDS Testr Calibration
This video shows how to calibrate the Oakton TDS pocket testers
Jan 27 2011 12:00AM
13
results (showing
1
-
13
)
Per Page
5
10
15
25
50
Close
Measuring pH of soil
View products by:
Job Title
|
Category
|
Brand
|
Part Number
|
Price
View product categories by:
Popularity
|
Favorites
|
Job Title
|
Price
|
Part Number
View popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Job Title
View reviews by:
Popularity
|
Product Title
|
Best Rated