United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Technical Resources
You are filtering by:
Tissue and Cell Culture
Clear all filters
Industries:
Fluid Handling
(4)
Industrial Manufacturing
(4)
Laboratory/Research
(7)
[+]
Show more
(3)
Life Science/Biotech
(13)
Masterflex Pumps
(4)
Pharmaceutical
(8)
[-]
Show less
Technical Resource Map
Looking for chemical compatibility ratings?
Use our searchable Chemical Compatibility Database
Need assistance?
Contact us
for
Expert Tech
Advice
Product
Selection
Custom
Solutions
Free Catalogs
Choose one for your specific needs.
Request your copy today.
Share
|
Home
>
Technical Resources
>
Tissue and Cell Culture
Technical Resource Library
Sort by
Best Match
Name
Recent Post Date
Search
Articles and White Papers
Sterilized Disposable Bioprocess Consumables
Disposable bioprocess containers have become extremely popular because of their extreme time and product cost savings.
Mar 21 2011 3:18PM
Articles and White Papers
Cole-Parmers Bioconnect Program
Sep 25 2009 4:31PM
Articles and White Papers
Cole-Parmers Bioconnect Program
Sep 25 2009 4:31PM
Articles and White Papers
Biotech Disposables
Explores the trend in biopharma of using disposables to limit staff costs and increase productivity
Sep 25 2009 4:31PM
Articles and White Papers
Cole-Parmers Bioconnect Program
Sep 25 2009 4:31PM
Articles and White Papers
Cole-Parmers Bioconnect Program
Details on Cole-Parmer's Bioconnect program; delivering custom disposable products and complete process validation.
Sep 25 2009 4:31PM
Articles and White Papers
Transwell Permeable Supports
General considerations and application chart for permeable supports
Aug 25 2006 10:26AM
Articles and White Papers
Poly-Lysine Substrates for Cell Culture
Poly-Lysine substrates promote firm attachment of neuronal cells and transfected cell lines in cell culture
Aug 25 2006 10:26AM
Articles and White Papers
Aspects of Nunc MAXISORP® MICROWELL® Certification
Details the certification procedure for adsorption testing and typical results of Cole-Parmer's Maxisorp Microwell plates
Aug 25 2006 10:26AM
Articles and White Papers
Integra CELLine
Highlights Integra's line of disposable plasticware for cell culture
Aug 25 2006 10:25AM
Articles and White Papers
Stability of Nunc-Immuno® Maxisorp® Surfaces
Describes the temperature influence on Nunc-Immuno MaxiSorp products stored for various periods of time.
Aug 25 2006 10:26AM
Articles and White Papers
MantaRay Single-useSpinner Flask Shows Excellent Cell Growth
Comparisons of cell growth in roller flasks, shake flasks, and the single-use MantaRay™ are discussed.
Aug 25 2006 10:26AM
Articles and White Papers
Principles in Adsorption to Polystyrene
The adsorption properties of Cole-Parmer's polystyrene well plates and the effects of surface chemistry
Aug 25 2006 10:26AM
Articles and White Papers
Negative Edge Effect in MICROWELL® ELISA
Discusses negative edge effect, i.e. unexpectedly lower optical densities in peripheral wells than in central wells of a MicroWell matrix, due to temperature difference
Aug 25 2006 10:26AM
14
results (showing
1
-
14
)
Per Page
5
10
15
25
50
Close
Oakton pHTestr Sensor Module Replacement
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title