Technical Resources
Post Date:
6/26/2013
Entry type:
Articles and White Papers
Topics:
Fluid Handling, Industrial Process, Water and Wastewater, Industrial Manufacturing, Fluid Handling
Tags: Flowmeters, Flowmeters
Flowmeters: What Type Will Work Best
for Your Application?
From magnetic to variable-area to Coriolis, the options in volumetric-based flowmeters are plentiful. Yet there are distinct differences in functionality, accuracy, and certainly price. To select the best model for your specific use without overspending, here’s a guide:
Coriolis Snapshot: Extremely high accuracy and no pressure drop. The meters track mass flow and offer a very high turn-down ratio. Yet initial expense is high, clogging can occur, and meters are larger in overall size.
Coriolis_Flowmeter.jpg
Coriolis meters offer true mass flow measurement through two designs: a single tube or two parallel tubes. They operate via
an oscillation which is induced in the tube(s) at a reference frequency. Based on Newton’s Second Law of Motion (F = m x a), the oscillation frequency will change with changes in mass flow rate.
Among the most accurate of technologies available, Coriolis flowmeters are suitable for a wide and growing range of gas and liquid applications. These devices provide multiparameter data on mass, density, and temperature. They are used in pharmaceutical manufacturing, wastewater treatment facilities, nuclear facilities, natural gas measurement and custody transfer.
Differential Pressure Snapshot: Very high accuracy, multiple calibrations, outputs and size. At the same time, this model can be used with water or gases only, and cannot handle particulates. It also requires power.
Differential Pressure flowmeters measure changes in pressure to determine flow velocity. They feature a flow-restrictive orifice or laminar flow element that evaluates the pressure drop through the restriction. The pressure drop between upstream and downstream points is proportional to the rate of flow. This technology works well when no moving parts are desired, or when an ultrafast response time is required.
Differential Pressure meters are typically found in more industrial applications, such as measuring the output of fuels (for example, benzines or jet fuel), in specialty chemical manufacturing, simple water measurement testing, or on aquafarms. They are also used in labs to measure and control the flow of gases when mixing them or separating them through chromatography.
Gear Snapshot: High accuracy; measurement is independent of fluid viscosity. No straight pipe runs required. Accuracy degrades slightly when measuring the flow of low-viscosity fluids.
Gear meters employ oval counter-synchronized rotors (gears) which are interlocked to rotate with the passing of liquid. The amount of fluid passing through the oval gears is well controlled, giving the meters a high level of accuracy. Designs are typically rugged and simple, allowing for installation in the most aggressive environments. In fact, gear meters are one of
a few types which are suited to high-viscosity fluids.
Used in hydraulics and other applications involving very
viscous liquids, gear meters work well in the pulp and paper industry, fuel or oil transfer, and manufacturing. Because the gears are stainless steel, they are ideal for the petrochemical industry or any application incorporating light to heavy oils. 
Magnetic Snapshot: No obstruction of flow path, no pressure drop, no moving parts. These meters can handle heavy slurries. Yet, the fluid measured must be conductive or water-based, and the meter must be grounded.
Magnetic meters (or magmeters) are available in two design styles: insertion and full-bore. Coils in the meter produce a magnetic field. When a conductive fluid is passed through the field, a voltage is produced through an electrode in the meter wall or insertion probe; this generated voltage is proportional
to the flow. Magmeters operate by measuring the electrical content of water or other fluids.
The magnetic technology contains no moving parts, and the full-bore designs offer no intrusions into the flow stream. Magmeters are higher-end flowmeters and are used in the
food & beverage industry, water purification, pulp and paper manufacturing, mining, chemical manufacturing, and petrochemical industries. They should not be used with low conductivity fluids such as
de-ionized water.
Paddle Wheel Snapshot: Fast response time. Easy maintenance. Inexpensive. Some may be difficult to install. Uses moving parts and requires a full pipe.
Paddle Wheel meters include those with rotating paddle wheels, propellers, and oscillating disks (multi-jet types). The rotating component is designed to provide a pulse when passing either a magnetic or optical sensor. The frequency of the pulses is proportional to the velocity of the fluid at one
point in the pipe or channel. These designs offer relatively high accuracy for their low cost. Some insertion versions are very easy to install while other styles are more difficult.
The paddle wheel style of flowmeter is often seen in rural
areas for irrigation, on aquafarms, in water/wastewater treatment, and for simple water measurement. These meters are also used in utilities and the oil and gas industries and can work with viscous fluids if a turbulent flow is present.
Gas Mass (Thermal Dispersion) Snapshot: No moving parts. Measures the mass of the gas, not the volume, so it is very accurate. However, the gas must be dry and free of particulates. Response time is fairly slow.
Gas Mass (Thermal Dispersion) meters operate with a side-stream flow of gas which is directed through a capillary. The capillary includes two external heater-sensor coils, one downstream from the other. Gas flow carries heat from the upstream coil to the downstream coil. The resultant temperature-dependent resistance differential at each coil is then measured. The gradient at the coils is linearly
proportional to the instantaneous flow rate.
With minimal invasiveness and no moving parts, these meters are used for chemical line monitoring, purging instrument air lines, and filtration loading. They may also control the flow in gas mixing and OEM applications.
Turbine Snapshot: High accuracy, millisecond response time, along with high pressure and temperature capabilities. Conversely, their moving parts can wear or become clogged. Not for low flow.
Turbine meters contain a bladed rotor positioned along the centerline of the flow stream. The rotating component is designed to provide a pulse when passing either a magnetic
or optical sensor. The frequency of the pulses is proportional
to the velocity of the fluid. Some designs offer high levels of accuracy and can handle slightly higher viscosity fluids than basic propeller-type designs. Also, some turbine designs
meet sanitary guidelines.
Irrigation and water purification are two common applications for turbine flowmeters. They are also used in the oil and gas, utilities, and wastewater industries. When used with sanitary connections, turbine meters control flow in food & beverage applications. These meters are not the best choice for low-flow applications.
Ultrasonic Snapshot: Very high accuracy. No pressure drop, no obstruction of flow path and no moving parts. Low maintenance costs. They are priced higher than some other technologies. Not a good choice for low-flow applications.
Portable_Doppler_Flowmeter.jpgUltrasonic meters offer more advanced technology and greater versatility than some other types. These designs measure the frequency shift of an ultrasonic
signal that is sent through the fluid. Two types of ultrasonic meters are Doppler and transit-time. Doppler technologies utilize
particles or aeration in the fluid as a reflective mechanism to gauge the velocity of the fluid. Transit-time technologies rely
on a frequency difference in forward and reverse signals sent though a clean liquid to gauge the velocity of the fluid; the
fluid must not have solids or aeration, as they will distort the sonic pulses. These are ideal technologies to create flow profiles through an existing process, when modifying piping
is not possible.
Because of their versatility, ultrasonic meters are used in a long list of industries, including facilities management, pulp and paper manufacturing, chemical manufacturing, and mining. Water/wastewater, petrochemical, and aquafarms also enlist this technology. Ultrasonic meters can be used to measure the corrosiveness of slurry fluid flow.
Variable Area Snapshot: Easy setup and use with low setup cost. Very low maintenance. Can be used for both liquids and gases. Yet, these meters provide low accuracy and may not withstand caustic media. No data output or recording capabilities.
Variable Area flowmeters are the veterans in the industry
and are the most commonly used. They are also referred to
as rotameters. Their design consists of a float—usually a sphere—enclosed in a tube. The float responds to change in velocity of the fluid (gas, air, or liquid) by moving up or down the flow tube. The variable area principle of operation is: fluid flow velocity raises a float in a tapered tube, increasing the
area for passage of the fluid. The greater the flow, the higher the float rises. The height of the float is directly proportional
to the flow rate. To determine flow, simply read the
graduated markings at the center of the float.
Variable area flowmeters can be used in laboratory and industrial applications, and when compared with other types
of flow instrumentation, are the most economical for indicating flow rate measurement when taking into account practicality and accuracy. These meters are used in academia in science labs for experimentation and education. They can be found in basic manufacturing including beverage manufacturing and chemical manufacturing as well as in water purification, aquafarms, and oil and gas applications. These meters should not be used with media that could coat the float or measuring tube.
Vortex Snapshot: Low to medium initial setup costs. Very low maintenance when used in clean flow conditions. Yet, the vortex meters may experience a low to medium pressure drop due to an obstruction in the flow path.
Vortex meters use a pressure sensor to measure the pressure pulses from vortices that come from the fluid passing a bluff body bar across the flow stream. A simple analogy of this phenomenon is that of a flag waving in the wind. The pulses
are proportional to the rate of flow. Many users find the technology appealing because it has no moving parts and
a low sensitivity to variations in process conditions.
Although many may be less familiar with the vortex flowmeter, it is an option that offers high accuracy. Because the meter body and vortex bar can be molded as one, the design is
ideal for use in either aggressive or high-purity applications. These meters work well in the oil and gas, water/wastewater, and food & beverage industries. They are also used in utilities, chemical manufacturing and water purification.
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title