log in or register
Gravity Feed Installation
of your Centrifugal Pump
This illustration shows a typical setup for a gravity feed system. Since most centrifugal pumps require priming, be sure to position pump at the bottom of the supply tank and retain a liquid level higher than the pump inlet port.
You can regulate the output flow with a ball valve or a flowmeter with a control valve on the discharge line.
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title