Technical Resources
Post Date:
8/25/2006
Entry type:
Articles and White Papers
Tags: Fittings, Tubing
Volume 4, Number 3, 2002
HOLD TIGHT!
  BarbLock™ Assemblies ensure leak-free tubing and fittings.
  • Easy to install locking mechanism—tubing and hose barb won't disengage.
  • Safe to use with pressures up to 140 psi (depending on tubing selected)
  • Secure tubing with no more crimps or distortion
  • Superior to other methods, including cable ties and hose clamps
    Do you have confidence in your tubing seals? You will now. Introducing BarbLock™ Assemblies, the most secure way to connect your Masterflex pump tubing to fittings, filters, and other barbed connections.
    The BarbLock system provides uniform 360º radial crimping to positively seal flexible tubing around fittings. These leak-free, durable seals are so strong and secure, they cannot be pulled or pushed off-even under pressure!
    A BarbLock assembly consists of three pieces: the sleeve, collet, and barbed fitting. To install the fitting, simply slide the sleeve and collet onto the tubing and then insert the barbed fitting into the tubing. Prelock the sleeve and collet by pressing them together until you hear a "click." Slide the sleeve and collet to the edge of the barbed fitting and lock with the BarbLock tool. The BarbLock assembly can then only be unlocked with the universal removal tool.
    Confidence? It's called BarbLock.
    BarbLock-TM TBM Corp
  • BarbLock assembly 30603-04
    Assembly tool 36603-12
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title