log in or register
NIST Atomic Fountain Clock
Gets Much Better with Time
Published with permission from NIST, photo copyright Geoffrey Wheeler Photography
NIST Atomic Fountain Clock NIST researchers (left to right) Steven Jefferts, Elizabeth Donley, and Tom Heavner with NIST F1, the world's best clock (as of Sept. 2005). The clock uses a fountain-like movement of cesium atoms to determine the length of the second so accurately that—if it were to run continuously—it would neither lose nor gain one second in 60 million years.
© 05 Geoffrey Wheeler Photography
The world’s best clock, NIST-F1, has been improved over the past few years and now measures time and frequency more than twice as accurately as it did in 1999 when first used as a national standard, physicists at the National Institute of Standards and Technology (NIST) report.
The improved version of NIST-F1 would neither gain nor lose one second in 60 million years, according to a paper published online Sept. 13 by the journal Metrologia.* NIST-F1 uses a fountain-like movement of cesium atoms to determine the length of the second. The clock measures the natural oscillations of the atoms to produce more than 9 billion "ticks" per second. These results then contribute to the international group of atomic clocks that define the official world time. NIST-F1 has been formally evaluated 15 times since 1999; in its record performance, it measured the second with an uncertainty of 0.53 x 10-15
The improved accuracy is due largely to three factors, according to Tom Parker, leader of the NIST atomic standards research group. First, better lasers, software and other components have made the entire NIST-F1 system much more reliable and able to operate for longer periods of time. Second, the atoms in the cesium vapor are now spread out over a much larger volume of space, reducing the frequency shifts caused by interactions among the atoms. (The formerly round cloud of atoms is now shaped like a short cigar.) Third, scientists are now better able to control magnetic fields within the clock and quantify the corrections needed to compensate for their effects on the atoms.
Improved time and frequency standards have many applications. For instance, ultraprecise clocks can be used to improve synchronization in precision navigation and positioning systems, telecommunications networks, and wireless and deep-space communications. Better frequency standards can be used to improve probes of magnetic and gravitational fields for security and medical applications, and to measure whether “fundamental constants” used in scientific research might be varying over time—a question that has enormous implications for understanding the origins and ultimate fate of the universe.
* T.P. Heavner, S.R. Jefferts, E.A. Donley, J.H. Shirley, T.E. Parker. 2005. NIST-F1: Recent improvements and accuracy evaluations. Metrologia (October 2005). Posted online Sept. 13.
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title