Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Technical Resources

Post Date:
6/22/2006

Entry type:
Case Studies

Topics:
Laboratory/Research, Masterflex Pumps, Pharmaceutical

Tags: Pumps - Masterflex® Peristaltic Pumps

 
Masterflex® Pharmaceutical Applications

Agar Dispensing Pump

Dispenses samples of agar from 0.05 to 8000 mL
Highly accurate performance: ±0.5%
No valves to clog
Easily sterilized
EASY-LOAD® II pump heads ensure low pulsation and accurate performance
Silicone tubing is recommended for purity


Pump product shown*
Pump system 77310-00, including:
Pump = 77200-62
Tubing = 96440-15
Drive = 77310-50


Also shown:*
14202-48 (nutrient agar medium)



View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title