Cleanroom Garments The correct order for cleanroom gowning: - Shoe covers
- Cap
- Hood
- Face mask
- Coveralls
- Boots
- Safety glasses
- Gloves
It is very important that garments be sized correctly. Improperly sized garments can raise the level of contaminants and cause safety concerns. Cleanroom garments must have these basic characteristics: - A minimum of seams. The seams must firmly envelop the edge of the material.
- Be loose fitting to eliminate abrasion
- Absence of pockets, belts, and pleated or tucked areas.
- Tight fabric weave to minimize the passage of dust.
- Fabric must reduce static electricity generation and still remain nonlinting.
- Garments should cover as much of the person as possible, especially around the neck area.
| |