Revco™ Ultra-low Temperature Freezer Selection Guide Ultima II Freezers Elite Freezers Value Series Freezers Accessories Freezer Specifications Ultima II-Series Control Panel - Ultima II models feature IntrLogic™ microprocessor control to monitor operation and alarm systems
- Available in -40 and -86°C models
- PEP (Pressure Equalization Port) on all Revco upright freezers relieves the inner chamber vacuum created under positive door seal when ambient air is introduced following door openings.
- Integrated door handle with latch has front accessible key lock; also accommodates a standard padlock
| | Ultima II |
IntrLogic microprocessor control system (panel shown below) sets, monitors, and regulates all freezer operating parameters with a touch of a button. Several times each second, a digital feedback loop surveys ambient temperature and refrigeration circuit temperature at critical points within the freezer. It then uses this information to regulate the freezer operation with minimal energy consumption. All Ultima II models include built-in voltage boost and surge suppressor, and eye-level controls.
| | Elite |
Elite Freezers include rugged cabinet construction, microprocessor controls, and alarm and monitor systems. Features audible and visible warnings for insufficient line voltage, low-battery condition, and the need to clean the filter.
| | Value series |
Value Series Freezers are a cost-effective alternative for a variety of industrial process and non-critical storage uses, the Revco Value series incorporates Legaci® refrigeration with Ultima II quality cabinet construction and reliable solid-state control for basic but dependable operation. All Revco ULT models feature Legacian air-cooled refrigeration system that uses heavy-duty, industrial grade compressors with strong pumping capacity and reserve cooling power. Freezers maintain set temperature even at an ambient temperature of 90°F (32°C) for reliable long-term performance.
Accessories for these freezers include temperature recorders, inventory racks and storage boxes, and grid dividers. Go to Top
| Features | Ultima II | Elite | Value | | General construction | Heavy gauge steel cabinet with epoxy coating to resist scratches and rusting. | • | • | • | Minimum 5" CFC-free foamed-in-place insulation (up to 3.5" in doors and lids) | • | • | • | | Recessed, heavy-duty aluminum casters for mobility | • | • | • | | Access port, 1" diameter for special accessories | • | • | • | | Temperature control system and electrical system | | Microprocessor-based or solid state controls | IntrLogic | Microproc | Solid state | Voltage boost system to detect and compensate for low voltage | • | • | • | | Surge suppressor to intercept voltage spikes | • | Optional* | Optional* | | Digital temperature display, adjustable in 1°C increments | • | • | • | | Key-operated main power switch to prevent tampering | • | • | • | | Alarm/monitor system | | Battery back up to operate alarm during power failure | | • | | | Battery capacity indicator | • | • | | | Touchpad alarm test for quick, accurate verification | • | • | | Audible and visual warnings in event of power failure or temperature deviation | • | • | Audible | Independent, tamper-resistant, adjustable high and low alarm setpoints (1°C increments) | • | • | | | RS-232 data port with system software | • | Optional* | | | Clean filter indicator light | • | • | | | Remote alarm contacts | • | • | • | | Refrigeration system | | Legaci refrigeration system | • | • | • | | 1-hp industrial compressors | • | • | • | | CFC-free refrigerant | • | • | • | | DuPont Suva® low-stage refrigerant | • | • | • | | Downfeed evaporator | • | • | • | | PEP (Pressure Equalization Port) | • | • | • | | Washable air-intake filter to protect condenser | • | • | • | Total warranty on compressor parts (including standard 2-year warranty on parts & labor)† | 5-yr | 4-yr | 3-yr | *Free standing
†USA and Canada only. Go to Top | | Panel features key-operated power and alarm switches to prevent tampering; alarm test to ensure alarm system is operating properly; touchpad controls to make setup easy; and warning lights to alert you to clean filter, change battery, or check voltage supply. | Go to Top | |