Technical Resources
Post Date:
8/25/2006
Entry type:
Articles and White Papers
Product Selection Guide
Topics:
Safety
Tags: Safety Products
Safety Gloves Sizing Guide
Glove Sizes
Hand Circumference Method
Hand Circumference Sizing Method
Measure the circumference of your hand around the palm area with fingers together and hand relaxed. This measurement in inches is closest to your actual glove size. For example, 7" is equal to a size 7 glove. Sizes may vary between different manufacturers and styles. Except where mentioned, gloves are sized according to men's hands. For easy size comparison, see table below.
Numerical size 6-7 7-8 8-9 9-10 11
General size XS S M L XL
Length
To determine the correct glove length you need, measure from the tip of the middle finger to where the cuff edge should be on the wrist or arm; then select the proper cuff and length.
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title