Technical Resources
Post Date:
8/25/2006
Entry type:
Conversions and Technical Data
Tags: Balances
Weighing Unit Conversion Table
To convert from grams to: Abbreviation multiply by
carats ct 5.0
dram 0.564383
grains gn 15.432358
momme mom 0.2666666
ounces oz 0.0352739
pennyweights dwt 0.6430149
pounds lb 0.0022046
scruple 0.7716179
tael (Hong Kong) tlh 0.0264554
tola (India) tol 0.0857353
tael (Singapore) tls 0.0264599
tael (Taiwan) tlt 0.0266666
troy ounces oz t 0.0321507
troy pounds lb t 0.0026792
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title