Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
LabGEN 700 homogenizer
LabGEN 700 Homogenizer is a versatile heavy-duty unit that can handle virtually any homogenizing task—dispersion, tissue processing, cell disruption, and preparing emulsions. The LabGEN 700 processes samples from 0.05 mL to 10 L with variable speeds from 10,000 to 30,000 rpm.