Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register
ProWeigh FIlters for TSS and VSS
Filters are composed of binderless borosilicate glass, have 1.5 µm porosity, and are processed thoroughly so you can start testing right out of the box! These VSS filters are vacuum rinsed three times with reagent grade water, dried, cooled, and desiccated to assure the best stability and repeatability.