Gear Pumps, Peristaltic Pumps, Laboratory Pumps—Pump Types and Definitions
What’s the difference between gear pumps, peristaltic pumps, and flexible impeller pumps? Can you define a progressing cavity? (Hint: It’s not something your dentist finds.)
Learn the various pump types and what they can do to in order to choose the right technology for your use. See how many of these you are familiar with:
BellowsThese pumps move fluid through a reciprocating bellows cavity that is coupled to a driving rod. These pumps are found in the “Metering” and “Positive Displacement” pump sections.
Centrifugal PumpsA rotating vaned disk attached to a drive shaft moves fluid without pulsation as it spins. The outlet can be restricted without damaging the pump.
Diaphragm Metering Pumps—Pulsations of one or two flexible diaphragms displace liquid while check valves control the direction of the fluid flow.
Standard Gear Pump System with Relief Valve, Calibration Column, and 3.0 to 7.0 GPM Flow Rate
Standard Gear Pump System with Relief Valve, Calibration Column, and 3.0 to 7.0 GPM Flow Rate
DrumA motor attached to an impeller is mounted on top of a drum. The impeller pushes fluid up the tube set when submersed.
Flexible Liner PumpsThe outer surface of an inert liner and inner surface of a rotating body block create a fluid channel used to gently pump fluids without pulsation.
Flexible ImpellerElastomeric impeller traps fluid between the impeller blades and a molded housing that sweeps fluid through the pump housing.
GearFluid is trapped between the teeth of two or three rotating gears. Gear pumps are good for high system pressure applications and are often magnetically driven.
HandManually powered pumps of various designs used for short term or repetitive liquid transfer applications.
Peristaltic Pumps/Peristaltic (tubing)—Fluid only contacts the tubing—rollers of a motor-driven pump head push the fluid along the tubing as they rotate. Noncontaminating and easy to clean.
PistonRotating pistons of varying stroke lengths pump fluids through check valves. Good for high-pressure applications.
Progressing CavityThe cavity between a screw-style rotor and stator force high-viscosity and particulate-filled fluids through the pump.
Rotary LobeLobe shaped rotors gently fill with and dispense fluids containing soft particles, solids, and highly viscous fluids. Sanitary construction is perfect for use in the food and beverage industries.
Rotary VaneOperate like the flexible impeller pumps but use an impeller made of a rigid material—useful for high-pressure or low-shear applications.
Syringe PumpsInfusion or withdrawal syringe pumps provide high-pressure and high accuracy for applications such as HPLC.
Submersible/ImmersibleCentrifugal pumps in a specialized housing used for emptying tanks or sumps.
4 Comments on "Gear Pumps, Peristaltic Pumps, Laboratory Pumps—Pump Types and Definitions"
  1. Great post I’ve ever seen. Actually I’m looking for this for a long time. I will run a shop very soon but don’t know about all kind of pumps. Now I know it very well. Thanks man.
    • Cole-Parmer Blog Team | December 2, 2020 at 4:27 pm | Reply
      Hi Andrew:
      Thank you for your comment. Glad we can help and good luck with your shop.
      Karen
  2. Swarnadip Ghosh | May 12, 2023 at 3:06 am | Reply
    I am looking for a peristaltic pump for setting up a microfluidic device. The pump should be connected to multiple inlets and maintain at least two different flow rates at a time.
    • Antylia Scientific Blog Team | May 16, 2023 at 11:46 am | Reply
      Hello Swarnadip:
      Please contact Avantor for Masterflex products. We no longer sell these products. Thank you!