CSI Revisited: Using Evidence Drying Cabinets
Evidence Drying Cabinet
Evidence Drying Cabinet
Crime scene investigators must gather evidence and make sure it dries safely without DNA cross-contamination. The Evidence Drying Cabinet provides a secure, ventilated chamber to dry this wet evidence, which may contain biohazards.
How does it work? An internal blower produces a flow of air to speed the drying process. HEPA and carbon filters filter the airflow exhaust and protect you from pathogens sometimes found in blood and body fluids. When air flow conditions are low, a filter alarm light alerts you to change the filter.
A key-activated UV light prevents DNA cross- contamination when transferring evidence into the cabinet. After the cabinet is disinfected and cleaned, the UV light activates to provide secondary decontamination by denaturing any remaining DNA left in the cabinet.
The cabinet’s interior floor is sloped to a drain to aid in cleanup. A safety glass door maximizes visibility and withstands UV exposure. The door may be secured with a user-supplied padlock or optional tamper-evidence tag to maintain chain of custody.
Be the first to comment on "CSI Revisited: Using Evidence Drying Cabinets"