Pittcon 2017 is just days away. Are you planning on attending? If so, join us in booth #2434 to experience the latest products designed to meet the challenges of laboratory science. Along with industry leading Masterflex® peristaltic pumps, highlights in the booth will include Ismatec® and Cole-Parmer® pumps as well as Jenway® spectrophotometers and PCRmax Alpha Cyclers.
These products are sure to create quite a buzz at Pittcon 2017:
- Cole-Parmer dual-drive syringe pump can be programmed to infuse each rack at different flow rates or to infuse and withdraw at the same time. A wide flow range from 0.0004 microliter/hr to 106.6 mL/min can be achieved.
- Ismatec Reglo Independent Channel Control (ICC) pumps allow individual control of flow and direction for each channel from the intuitive keypad or a computer. The ICC pump delivers continuous pumping or precision dispensing up to 43 mL/min.
- Jenway 7205 Scanning UV/Visible spectrophotometer with diode array technology is extremely fast, reliable and virtually maintenance free. The multi-element detector on this scanning spectrophotometer measures the entire wavelength range at once, reducing wait times.
- PCRmax Alpha Cycler 2 condenses two independent blocks into one compact system, allowing for two different protocols to run at the same time. A Program Wizard generates a protocol specific to your sequence in seconds—a quick way to optimize new assays.
Be sure to stop by booth #2434 at Pittcon 2017 for more information about these and our comprehensive selection of products for the laboratory research community.
Be the first to comment on "Cole-Parmer Will Be Exhibiting at Pittcon 2017"