The Oakton EcoTestr pH2+ pocket meter video presents the newest addition to the Oakton product line. The new pocket meter features updates to make measuring water quality easier and faster. The display is larger with a better viewing angle, has indicators for battery life, readiness, and calibration, and shows both the parameter and temperature readings at the same time. Learn more about this cool tool by watching the video below:
Be the first to comment on "Water You Waiting For? Watch the Oakton EcoTestr ph2+ Pocket Meter Video"