PCRmax Reagent Kits: Largest Offering of Lyophilized qPCR Detection Kits Available on the Market
qPCR detection kits
PCRmax qPCR Detection Kits provide the largest offering of lyophilized qPCR detection kits available on the market today. With more than 800 available, the kits offer convenience, accurate and quick results, and exceptional value at 150 tests per kit.
All contents of the kit are lyophilized for simple transport and storage, no refrigeration required. Each kit uses the speed and sensitivity of qPCR to obtain the most accurate data quickly. Each kit is also designed to ensure the broadest detection profile and detection of all clinically relevant strains and subtypes. Positive copy number standard curve provides accurate quantification.
Kits are available with or without mastermix. Detection kits with mastermix contain all the necessary components for fast and accurate results. To use your own mastermix, order kits without mastermix.
Look for these kits and more:
Systemic Viral Pathogen Detection Kits – Prevent economic loss, especially in the dairy and agricultural industry, using these viral detection kits to accurately assess whether these disease-causing pathogens are present.
Human Viral Pathogen Detection Kits – Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes.
Human Microbial Pathogen Detection Kits – Quickly and accurately detect the presence of human microbial pathogens.
Plant and Animal Allergen and Speciation Detection Kits – No need to refrigerate—all contents are lyophilized for simple transport and storage.
Genetically Modified Organism Detection Kits – Screen for genetically modified organisms (GMOs) in food, feed, and seeds.
For more information on the PCRmax reagent kits, call our product specialists at 1-800-323-4340.
Be the first to comment on "PCRmax Reagent Kits: Largest Offering of Lyophilized qPCR Detection Kits Available on the Market"