Plastic vs. Glass Carboys
Carboys are typically used to store liquids, chemicals, solvents, or corrosive fluids. Often used in labs and industry, sometimes they are chosen for home brewing uses as well.
Plastic or Glass Carboys?
The question becomes, depending on your specific application, which is better—plastic or glass?When choosing a carboy, keep in mind chemical compatibility and FDA-compliance, if needed. Capacities vary. Choose the largest capacity you are likely to need (or order multiples to reach that capacity).
 Advantages of Plastic
 Disadvantages of Plastic
Scratch easily
Tough to clean
Generally used
 Advantages of Glass
 Disadvantages of Glass
Easier to clean
Less prone to contamination
Heavy to carry
Carboy styles include square/rectangular, UN-Approved, round, and heavy-duty vacuum that withstand full vacuum applications. Available accessories include filling/venting caps, racks, faucets, and replacement spigots.
Need help selecting a carboy for your application? Contact our product experts at 1-800-323-4340.
Be the first to comment on "Plastic vs. Glass Carboys"