New Environmental Express® HotBlock® 200 System Takes Metals Digestion Up a Notch
Environmental Express HotBlock™ 200 Digestion Heating Blocks
New Heating Blocks System for Metals Digestion
Here’s a great product if you work with metals digestion. This new HotBlock 200 system allows you to perform metals digestion at higher temperatures—up to a maximum 200°C (392°F), which is above industry standards.
The HotBlock 200 system is a redesign of the HotBlock Pro with new features but keeps the same dependable graphite block and Kydex® housing as the company continues to use metals-free materials in its construction to avoid cross-contamination and provide high chemical resistance. These units are available from 25 to 96 wells and accept cup sizes of 15 mL, 50 mL, and 100 mL. Custom units are also available.
The HotBlock 200 system also features redesigned controllers. The easy-to-use external controllers feature automatic shut-off, alarms, and ability to save profiles with ramp and soak settings. The dual-block controller can control up to two heating blocks independently and simultaneously. They can be safely mounted to the outside of a fume hood or placed on a benchtop.
Need more information on the HotBlock 200 metals digestion system? Call our technical experts at 1-800-343-5319.
Be the first to comment on "New Environmental Express® HotBlock® 200 System Takes Metals Digestion Up a Notch"