Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 229
General-Purpose Acrylic in-Line Flowmeter for Liquid
to
Male Pipe Adapter
Image
Product Detail
SKU
General-Purpose Acrylic In-Line Flowmeter for Liquids; 2 to 20 GPM, 1" (F-41000LN-16)
Category: Variable Area Flowmeters
Description: Easy-View Acrylic In-Line Flowmeters
Brand:
Cole-Parmer
Model Number:
F-41000LN-16
YO-32477-30
More Details
Cole-Parmer Spring-Loaded Inline Flowmeter for Water, Air or Nitrogen; (YO-32211-16)
Category: Variable Area Flowmeters
Description: Cole-Parmer Economical Spring-Loaded Flowmeters
Brand:
Cole-Parmer
YO-32211-16
More Details
External antenna with magnet for Oakton Wireless Datalogger system. Us (YO-35710-30)
Category: General Accessories
Description: General Accessories and Replacement Items
Brand:
Oakton
YO-35710-30
More Details
CoorsTek Wide-Form Crucible, Porcelain; 250 mL, 102 mm top OD, 60 mm H (YO-17804-14)
Category: Crucibles
Description: CoorsTek Wide-Form Crucibles, Porcelain
Brand:
CoorsTek
YO-17804-14
More Details
Adjustable Safety Shield, 12" x 12" (8550-255)
Category: Safety Shields
Description: Adjustable Safety Shields
Brand:
Cole-Parmer
Model Number:
8550-255
YO-41052-03
More Details
Male Pipe Adapter, StainlessSteel, 1/8" x 1/4", 1 PACK (YO-30621-33)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30621-33
More Details
6846
results
<<<Previous
225
226
227
228
229
View by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
Page:
1
130
140
150
160
170
180
190
199
200
201
202
203
204
205
206
207
208
209
210
211
212
213
214
215
216
217
218
219
220
221
222
223
224
225
226
227
228
229
General-Purpose Acrylic in-Line Flowmeter for Liquid to Male Pipe Adapter
>
31125329-ts-toc_2 31125329tstoc2