Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 229
8-Port Manifold (Yo-24802-00)
to
983
Image
Product Detail
SKU
8-port manifold (YO-24802-00)
Category: Liquid Handling System
Description: 8-Port Manifold
Brand:
Cole-Parmer
YO-24802-00
More Details
83l Benchtop Heated Shaking Incubator - 120vac / 60hz
Category: Incubating Shakers
Description: Lab Companion Digital Incubating/Refrigerating Shakers
Brand:
Lab Companion
YO-79510-48
More Details
83l Refrigerated Benchtop Shaking Incubator - 120vac / 60hz
Category: Incubating Shakers
Description: Lab Companion Digital Incubating/Refrigerating Shakers
Brand:
Lab Companion
YO-79510-54
More Details
83l Refrigerated Benchtop Shaking Incubator - 230 VAC / 50hz
Category: Incubating Shakers
Description: Lab Companion Digital Incubating/Refrigerating Shakers
Brand:
Lab Companion
YO-79510-52
More Details
9 1/2" wide flexible drum heater, 55 gallon, 1500 watt, 240 VAC, 70-19 (YO-09203-32)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09203-32
More Details
983 : Particle Counter w/ Six Particle D (FLUKE-983)
Category: Air Sampling
Description: Fluke Particle Counter
Brand:
Fluke
Model Number:
FLUKE-983
YO-10101-58
More Details
6846
results
<<<Previous
225
226
227
228
229
View by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
Page:
1
130
140
150
160
170
180
190
199
200
201
202
203
204
205
206
207
208
209
210
211
212
213
214
215
216
217
218
219
220
221
222
223
224
225
226
227
228
229
8-Port Manifold (Yo-24802-00) to 983
>
31125329-ts-toc_2 31125329tstoc2