1 Ml Syringe without Needle

Image
  Product Detail
SKU

Gastight High-Performance Syringes; capacity, 1.0mL; gauge, 22; syringe type, cemented-needle

Category: Hamilton Gastight High-Performance Syringes
Description: Hamilton Gastight High-Performance Syringes with Cemented Needle
Brand: Hamilton


Gastight High-Performance Syringes; capacity, 1 mL; gauge, 22; syringe type; removable-needle

Category: Hamilton Gastight High-Performance Syringes
Description: Hamilton Gastight High-Performance Syringes with Removable Needle
Brand: Hamilton


Hamilton Gastight Syringes with luer tip; capacity, 1.0mL, 22 gauge needle

Category: Hamilton Gastight High-Performance Syringes
Description: Hamilton Gastight High Performace Syringes with Luer Tip
Brand: Hamilton


Syringes with PTFE luer lock; syringe capacity; 1 mL; needle gauge; 22 (81320)

Category: Hamilton Gastight High-Performance Syringes
Description: Hamilton Gastight Syringes with PTFE Luer Lock
Brand: Hamilton




1 Ml Syringe without Needle
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended |  Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended

Previous: Hamilton Gastight High-Performance Syringes with Removable Needle  |  Next: Syringe Types