1 Mm Id Capillary Tube

Image
  Product Detail
SKU

Replacement capillary tube with scale

Category: Accesories for Tensiometers and Surface Tension Apparatus
Description: Replacement Glass Capillary Tube for Surface Tension Apparatus
Brand: Lab Products


Rotor Microtube HOLDS 24 CAPILLARY TUBES

Category: Microcentrifuges
Description: Accessories for Thermo Scientific Heraeus Pico and Fresco Microcentrifuges
Brand: Thermo Scientific


Cole-Parmer Aluminum Blocks, for 12 x 15/16mm ID Test Tubes

Category: Chilling & Heating Blocks
Description: Cole-Parmer Benchtop Chilling/Heating/Mixing Blocks Accessories
Brand: Cole-Parmer


Cole-Parmer Aluminum Blocks, for 20 x 12/13mm ID Test Tubes

Category: Chilling & Heating Blocks
Description: Cole-Parmer Benchtop Chilling/Heating/Mixing Blocks Accessories
Brand: Cole-Parmer


Capillary tubes; 1.5-mm; 1000/pack (AT4042)

Description: General Accessories
Brand: Lab Products


Capillary tubes; 2.0-mm; 1000/pack (AT4043)

Description: General Accessories
Brand: Lab Products


Glass capillary tubes, open one end, pack of 100

Category: Digital Melting Point Apparatuses
Description: Stuart Digital Melting Point Apparatus
Brand: Cole-Parmer


Glass capillary tubes, open both ends, pack of 100

Category: Melting Point
Description: Stuart Analog Melting Point Apparatus
Brand: Cole-Parmer


Glass capillary tubes, closed at both ends, pack of 100

Category: Melting Point
Description: Stuart Analog Melting Point Apparatus
Brand: Cole-Parmer


Capillary tubes, 1.3 to 1.5-mm dia., 150/pk.

Category: Melting Point
Description: METTLER TOLEDO Digital Melting Point Apparatus
Brand: Mettler Toledo


Inoculating needle and loop; color, blue; dimensions; 9"L x 1 mm ID loop; 960/box

Category: Essentials and Accessories for Microbial Analysis
Description: Inoculating Needle and Loops
Brand: Cole-Parmer


Inoculating needle/loop, 1-µL cap, 1-mm ID loop x 7-3/4"L, clear

Category: Inoculation Loops and Needles
Description: Combination Inoculating Needle/Loop
Brand: Cole-Parmer


Solenoid Valve, 2-Way Normall Closed, 24V, with 1/8" Tube Connections, EPDM diaphragm

Category: Solenoid Valves
Description: Compact Direct Operated 2- and 3-Port Solenoid Valve for Chemicals
Brand: SMC Industries


Black Rubber Vacuum Tubing, 1/8" ID x 3/8" OD, 25ft. (YO-96143-01)

Category: Rubber Tubing
Description: Black Rubber Vacuum Tubing
Brand: Vacuum tubing


Black Rubber Vacuum Tubing, 3/16" ID x 9/16" OD, 25ft. (YO-96143-02)

Category: Rubber Tubing
Description: Black Rubber Vacuum Tubing
Brand: Vacuum tubing


Black Rubber Vacuum Tubing, 1/4" ID x 1/2" OD, 25ft. (YO-96143-04)

Category: Rubber Tubing
Description: Black Rubber Vacuum Tubing
Brand: Vacuum tubing


Black Rubber Vacuum Tubing, 1/4" ID x 5/8" OD, 25ft. (YO-96143-06)

Category: Rubber Tubing
Description: Black Rubber Vacuum Tubing
Brand: Vacuum tubing


Black Rubber Vacuum Tubing, 3/8" ID x 7/8" OD, 25ft. (YO-96143-08)

Category: Rubber Tubing
Description: Black Rubber Vacuum Tubing
Brand: Vacuum tubing


Black Rubber Vacuum Tubing, 1/2" ID x 1" OD, 25ft. (YO-96143-10)

Category: Rubber Tubing
Description: Black Rubber Vacuum Tubing
Brand: Vacuum tubing


Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft. (YO-96143-12)

Category: Rubber Tubing
Description: Black Rubber Vacuum Tubing
Brand: Vacuum tubing




1 Mm Id Capillary Tube
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended |  Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended

Previous: Vacuum Tubing  |  Next: 50 Um Id Capillary Tubes