Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
1000 Pound Portable Digital Scale
Image
Product Detail
SKU
A&D HLWP Portion Control Washdown Scale, 1000 G X 0.5 G
Category: A&D Weighing Food Portion Scales
Description: A&D Weighing HLWP Portable Washdown Scales
Brand:
A&D Weighing
Model Number:
HL-1000WP
YO-11114-82
More Details
A&D Portable Stainless Steel Washdown Scale, 1000 g (SK-1000WP)
Category: A&D Weighing Food Portion Scales
Description: A&D Weighing SK Stainless Steel Washdown Scales
Brand:
A&D Weighing
Model Number:
SK-1000WP
YO-11114-00
More Details
Adam Highland Portable Balance, 1000g X 0.01g
Category: Adam Equipment Portable Balances
Description: Adam Equipment Highland Portable Balances
Brand:
Adam Equipment
YO-11712-19
More Details
Adam Highland Portable Balance, 1000g X 0.01g (220 VAC)
Category: Adam Equipment Portable Balances
Description: Adam Equipment Highland Portable Balances
Brand:
Adam Equipment
YO-11712-27
More Details
A&D SJ Series Digital Portion Scale, 1000 G X 0.5 G
Category: A&D Weighing Food Portion Scales
Description: A&D Weighing SJ Portion Scales
Brand:
A&D Weighing
Model Number:
SJ-1000HS
YO-11111-21
More Details
Ambrobe AM-270 Digital Multimeter, 1000 V
Category: Amprobe Digital Multimeters
Description: Amprobe 200 Series Digital Multimeter
Brand:
Amprobe
Model Number:
AM-270
YO-20037-31
More Details
1000 Pound Portable Digital Scale
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Cole-Parmer Symmetry
|
Next: Adam Equipment Highland Portable Balances
>
31125329-s-content_2 31125329scontent2