Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
15 Gallon Plastic Drum
Image
Product Detail
SKU
Cone bottom tank, 15 gallons, 45° cone (YO-06960-26)
Category: Plastic Tanks and Drums
Description: Cone Bottom Tanks
Brand:
Cole-Parmer
YO-06960-26
More Details
3" wide flexible drum heater, 15 gallon, 500 watt, 120 VAC, 70-425deg (YO-09200-10)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09200-10
More Details
9 1/2" wide flexible drum heater, 15 gallon, 1000 watt, 120 VAC, 70-19 (YO-09203-10)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09203-10
More Details
15-gallon flexible silicone rubber drum heater with thermostat: 50 to (DHCS21)
Category: Drum Heaters
Description: BriskHeat Heavy-Duty Flexible Drum Heaters with Thermostat
Brand:
BriskHeat
Model Number:
DHCS21
YO-03106-70
More Details
15-gallon flexible silicone rubber drum heater with thermostat: 50 to (DPCS11)
Category: Drum Heaters
Description: BriskHeat Heavy-Duty Flexible Drum Heaters with Thermostat
Brand:
BriskHeat
Model Number:
DPCS11
YO-03106-74
More Details
15-gallon flexible silicone rubber drum heater with thermostat: 50 to (DPCS21)
Category: Drum Heaters
Description: BriskHeat Heavy-Duty Flexible Drum Heaters with Thermostat
Brand:
BriskHeat
Model Number:
DPCS21
YO-03106-78
More Details
15-gallon flexible silicone rubber drum heater with thermostat: 50 to (DHCS11)
Category: Drum Heaters
Description: BriskHeat Heavy-Duty Flexible Drum Heaters with Thermostat
Brand:
BriskHeat
Model Number:
DHCS11
YO-03106-66
More Details
Drip Sentinel Drum-top Workstation For 55 Gallon Steel Drums
Category: Drum Pump Accessories
Description: Drip Sentinel Drum-Top Workstation
Brand:
Drip Sentinel
Model Number:
3200-BK
YO-79012-99
More Details
Justrite Drum Lock, Plastic Drum Set (08509)
Category: Storage and Handling
Description: Justrite
®
Drum Locks
Brand:
Justrite
Model Number:
08509
YO-81792-65
More Details
Tank; PE, 15 gallons. Includes ½" PVC bulkhead fitting.
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
J40496
YO-74113-93
More Details
Tank; PE, 15 gallons. Includes 3/8" PVC bulkhead fitting
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
J40492
YO-74113-95
More Details
15 Gallon tank for mechanical pumps
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
STS15NC
YO-74208-90
More Details
15 Gallon tank for digital pumps
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
STS15N-02
YO-74208-94
More Details
Portable Pressurized Eyewash with Drench Hose, 15 gallon
Category: Portable/Self-Contained Eyewash
Description: Bradley Portable Pressurized Eyewash with Drench Hose
Brand:
Bradley
Model Number:
S19-788
YO-86000-12
More Details
Plastic drum dolly (YO-06341-62)
Category: Thermo Scientific HyClone HyQ 3-D Single Web BioProcess containers
Description: 5 ports, bottom dispense
Brand:
HyClone
YO-06341-62
More Details
Rectangular Mobile Tub, LDPE, 15 gallon
Category: Mobile Bins and Tubs
Description: Rectangular Mobile Tubs, LDPE
Brand:
Cole-Parmer
YO-06745-20
More Details
Stand for 15 gallon tank 06960-26 (48900)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
48900
YO-06960-88
More Details
High-density polyethylene tap faucet (COM4411)
Category: Bottle Accessories
Description: Plastic Drum Faucets
Brand:
Guaranteed In Stock Program
Model Number:
COM4411
YO-06061-03
More Details
Thermo Scientific Nalgene needle-type tank spigot, 3/4" NPT(M)
Category: Bottle Accessories
Description: Plastic Drum Faucets
Brand:
Thermo Scientific Nalgene
Model Number:
6420-0750
YO-06070-30
More Details
Faucet for 06065-70, 5/pk (FLO-RITE)
Category: Bottle Accessories
Description: Plastic Drum Faucets
Brand:
Cole-Parmer
Model Number:
FLO-RITE
YO-06078-00
More Details
15 Gallon Plastic Drum
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Thermo Scientific Replacements Accessories
|
Next: Bottles Accessories
>
31125329-s-content_2 31125329scontent2