Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
150 Psi Micro Water Pump
Image
Product Detail
SKU
High Precision Micro Metering Pump, 500 psi, programmable controller
Category: Piston Pumps
Description: Micro Metering Pump
Brand:
Cole-Parmer
Model Number:
120936
YO-74930-70
More Details
Mini Lubricator with bowl guard, Max 32 SCFM, Max Pressure of 150 PSI,
Category: Accessories for Compressors - Air Equipment
Description: ARO-Flo Series Filters, Regulators, and Lubricators
Brand:
ARO-Flo
Model Number:
L36121-100
YO-75300-66
More Details
Compact Air Filter, 5 micron filter, Max 73 SCFM, Max Pressure 150 PSI
Category: Accessories for Compressors - Air Equipment
Description: ARO-Flo Series Filters, Regulators, and Lubricators
Brand:
ARO-Flo
Model Number:
F35221-400
YO-75300-80
More Details
Thermo Scientific Nalgene Aspirator pump, 7.5 psi minimum water pressu
Category: General Accessories
Description: Replacement Accessories
Brand:
Nalgene
Model Number:
6140-0010
YO-06378-30
More Details
Cole-Parmer Inert Solenoid Self Priming Micro Pumps, 150 microL, 12 VD
Category: Diaphragm Pumps
Description: Cole-Parmer Self-Priming Micro Pumps
Brand:
Cole-Parmer
Model Number:
150SP12150-4EE
YO-73120-54
More Details
Cole-Parmer Inert Solenoid Self Priming Micro Pumps, 150 microL, 24 VD
Category: Diaphragm Pumps
Description: Cole-Parmer Self-Priming Micro Pumps
Brand:
Cole-Parmer
Model Number:
150SP24150-4EE
YO-73120-56
More Details
Microannular Gear Pump, 5microliter to 255ml/min, 116 PSI with analog
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24689
YO-73100-00
More Details
Microannular Gear Pump, 1.2 microliter to 72ml/min, 73 PSI with analog
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24690
YO-73100-02
More Details
Microannular Gear Pump, 0.15microliter to 9ml/min, 22 PSI with analog
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24692 (H11010103 MZR 2521/H66020300 MZR SK-KG-21)
YO-73100-06
More Details
Microannular Gear Pump, 5microliter to 255ml/min, 116 PSI with digital
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24693
YO-73100-10
More Details
Microannular Gear Pump, 1.2 microliter to 72ml/min, 73 PSI with digita
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24694
YO-73100-12
More Details
Microannular Gear Pump, 0.15microliter to 9ml/min, 22 PSI with digital
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24696
YO-73100-16
More Details
Microannular Gear Pump, 5microliter to 255ml/min, 1160 PSI with digita
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24697
YO-73100-50
More Details
Microannular Gear Pump, 1.2 microliter to 72ml/min, 435 PSI with digit
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24698
YO-73100-52
More Details
Microannular Gear Pump, 0.3microliter to 18ml/min, 145 PSI with digita
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24699
YO-73100-54
More Details
Microannular Gear Pump, 0.3microliter to 18ml/min, 44 PSI with analog
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24691
YO-73100-04
More Details
PTFE Pulse Dampeners With PTFE Diaphragm, 3/8" NPT
Category: Accessories for Positive Displacement Pumps
Description: Pulse Dampeners
Brand:
PTFE
Model Number:
AD-10TT (3/8 in NPT)
YO-74021-00
More Details
Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
Category: Accessories for Positive Displacement Pumps
Description: Pulse Dampeners
Brand:
Santoprene
Model Number:
853327;AD-10PS
YO-74021-04
More Details
PTFE Pulse Dampeners With PTFE Diaphragm, 3/4" NPT
Category: Accessories for Positive Displacement Pumps
Description: Pulse Dampeners
Brand:
PTFE
Model Number:
AD-25TT (0.75 in NPT)
YO-74021-10
More Details
Polypropylene Pulse Dampeners With Santoprene Diaphragm, 1" NPT
Category: Accessories for Positive Displacement Pumps
Description: Pulse Dampeners
Brand:
Santoprene
Model Number:
853348;AD-25PS
YO-74021-14
More Details
150 Psi Micro Water Pump
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Liquid Micro Flow Meter
|
Next: Micro Metering
>
31125329-s-content_2 31125329scontent2