Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
18 High Temperature Gloves
Image
Product Detail
SKU
Zetex High-Temperature Gloves, up to 1100°F, one size fits all, 14
Category: Temperature-Resistant Gloves
Description: Zetex High-Temperature Gloves and Finger Cots
Brand:
Zetex
Model Number:
ZG1
YO-86364-00
More Details
ZetexPlus High-Temperature Gloves, up to 2000°F, one size fits all
Category: Temperature-Resistant Gloves
Description: ZetexPlus High-Temperature Gloves
Brand:
ZetexPlus
Model Number:
16000-14-50-00
YO-86364-10
More Details
Zetex High-Temperature Gloves with Aluminized Back, up to 1100°F,
Category: Temperature-Resistant Gloves
Description: Zetex High-Temperature Gloves and Finger Cots
Brand:
Zetex
Model Number:
160001-14-00-00
YO-86364-20
More Details
Zetex High-Temperature Finger Cots, up to 1100°F, one size fits al
Category: Temperature-Resistant Gloves
Description: Zetex High-Temperature Gloves and Finger Cots
Brand:
Zetex
Model Number:
ZFC
YO-86364-02
More Details
Whirl-Pak 18 oz High Temperature Bags, 500/box
Category: Whirl-Pak Sterile Sampling Bags
Description: Whirl-Pak high temperature bag
Brand:
Whirl-Pak
YO-06498-16
More Details
QRP™ Extreme-Temperature Gloves, 18"L, Large
Category: Cleanroom and Static Control Gloves
Description: QRP Extreme-Temperature Gloves
Brand:
QRP Incorporated
Model Number:
55G
YO-33689-22
More Details
NIST-Traceable Calibration; Single High-Temperature Dry-Well Calibrato (YO-17002-18)
Category: Temperature Calibrations
Description: Temperature Calibrator Calibrations
Brand:
Cole-Parmer
YO-17002-18
More Details
Horizontal Tube Furnace, 18-1/2" Tube Length, 1800°C, 208 / 240 V (RHTH 120/150/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/150/18/C42
YO-33334-39
More Details
Vertical Tube Furnace, 18-7/8" Tube Length, 1800°C, 208 / 240 VAC (RHTV 120/150/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/150/18/C42
YO-33334-48
More Details
Horizontal Tube Furnace, 24-3/8" Tube Length, 1800°C, 208 / 240 V (RHTH 120/300/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/300/18/C42
YO-33334-42
More Details
Horizontal Tube Furnace, 36-1/4" Tube Length, 1800°C, 208 / 240 V (RHTH 120/600/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/600/18/C42
YO-33334-45
More Details
Vertical Tube Furnace, 24-3/4" Tube Length, 1800°C, 208 / 240 VAC (RHTV 120/300/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/300/18/C42
YO-33334-51
More Details
Vertical Tube Furnace, 34-5/8" Tube Length, 1800°C, 208 / 240 VAC (RHTV 120/600/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/600/18/C42
YO-33334-54
More Details
Horizontal Tube Furnace, 18-1/2" Tube Length, 1600°C, 208 / 240 V (RHTH 120/150/16/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/150/16/C42
YO-33334-37
More Details
Horizontal Tube Furnace, 18-1/2" Tube Length, 1700°C, 208 / 240 V (RHTH 120/150/17/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/150/17/C42
YO-33334-38
More Details
Vertical Tube Furnace, 18-7/8" Tube Length, 1600°C, 208 / 240 VAC (RHTV 120/150/16/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/150/16/C42
YO-33334-46
More Details
Vertical Tube Furnace, 18-7/8" Tube Length, 1700°C, 208 / 240 VAC (RHTV 120/150/17/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/150/17/C42
YO-33334-47
More Details
Cole-Parmer 150-mm Correlated Flowmeter;Aluminum with 316 SS float; Fo (YO-03227-18)
Category: Cole-Parmer 150-mm Correlated Flowmeters
Description: Correlated Flowmeters with High-Resolution Valves
Brand:
Cole-Parmer
YO-03227-18
More Details
Cole-Parmer 150-mm Correlated Flowmeter;Brass with 316 SS float; For L (YO-03295-18)
Category: Cole-Parmer 150-mm Correlated Flowmeters
Description: Correlated Flowmeters with High-Resolution Valves
Brand:
Cole-Parmer
YO-03295-18
More Details
Cole-Parmer 65-mm Correlated Flowmeter , Aluminum with SS float; For (YO-32044-18)
Category: Cole-Parmer 65-mm Correlated Flowmeters
Description: Correlated Flowmeters with High-Resolution Valves
Brand:
Cole-Parmer
YO-32044-18
More Details
18 High Temperature Gloves
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Nabertherm Mini Tube Furnaces
|
Next: Low Temperature Tube Furnace
>
31125329-s-content_2 31125329scontent2