Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Related Categories
5000 Ml Erlenmeyer Flask
3000 Ml Erlenmeyer Flask
Disposable Erlenmeyer Flask
Sterile Erlenmeyer Flasks Plastic
Erlenmeyer 250 Ml
Nalgene Fep Erlenmeyer Flask
Erlenmeyer Flasks Polypropylene 5000ml
Spinner Flask 250 Ml
Thermo Scientific Nalgene Polypropylene Erlenmeyer Flasks
100 Ml Plastic Erlenmeyer Flask
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
250 Ml Erlenmeyer Flask
Image
Product Detail
SKU
Corning 250 mL PC Flask w/vented cap, 50/CS
Category: Plastic Erlenmeyer Flasks
Description: Corning Sterile Disposable Erlenmeyer Flasks, PC
Brand:
Corning
YO-06114-62
More Details
Universal 250-ml Erlenmeyer flask clamp
Category: Shakers
Description: Accessories for Shakers
Brand:
Lab Products
Model Number:
30154BI
YO-51706-80
More Details
Thermo Scientific Low Cost Orbital Shaker for 250 mL Erlenmeyer flasks (2346Q)
Category: Orbital Shakers
Description: Thermo Scientific Orbital Shaker
Brand:
Thermo Scientific
Model Number:
2346Q
YO-04730-24
More Details
Thermo Scientific Low Cost Orbital Shaker for 250 mL Erlenmeyer flasks (2346-1CEQ)
Category: Orbital Shakers
Description: Thermo Scientific Orbital Shaker
Brand:
Thermo Scientific
Model Number:
2346-1CEQ
YO-04730-26
More Details
Thermo Scientific Nalgene polypropylene Erlenmeyer flask, 250 mL
Category: Plastic Erlenmeyer Flasks
Description: Thermo Scientific Nalgene Polypropylene Erlenmeyer Flasks
Brand:
Thermo Scientific Nalgene
YO-06109-30
More Details
PYREX Brand 4995 flask; 250 mL, case of 6
Category: Glass Erlenmeyer Flasks
Description: PYREX Brand 4995 Erlenmeyer Glass Flasks
Brand:
Pyrex
YO-34555-21
More Details
Pyrex Brand 5100 flask; 250 mL, pack of 12
Category: Glass Erlenmeyer Flasks
Description: Pyrex Brand 5100 Erlenmeyer Glass Flasks
Brand:
Pyrex
YO-34556-02
More Details
PYREX Brand 4980 flask; 250 mL, pack of 12
Category: Glass Erlenmeyer Flasks
Description: PYREX Brand 4980 Glass Flasks
Brand:
Pyrex
YO-34503-05
More Details
PYREX Brand 4980 flask; 250 mL, case of 48
Category: Glass Erlenmeyer Flasks
Description: PYREX Brand 4980 Glass Flasks
Brand:
Pyrex
YO-34503-25
More Details
KIMAX flask, 250 mL, 48/CS
Category: Glass Erlenmeyer Flasks
Description: Kimax Glass Flasks
Brand:
Kimax
YO-34524-16
More Details
PYREX VISTA™ narrow mouth flask, 250 mL, 24/CS
Category: Glass Erlenmeyer Flasks
Description: PYREX VISTA Glass Flasks
Brand:
Pyrex
YO-34530-86
More Details
PYREX Brand 4985 flask with screw cap; 250 mL, pack of 6
Category: Glass Erlenmeyer Flasks
Description: PYREX Brand 4985 Glass Flasks
Brand:
Pyrex
YO-34554-03
More Details
PYREX Brand 4985 flask with screw cap; 250 mL, case of 24
Category: Glass Erlenmeyer Flasks
Description: PYREX Brand 4985 Glass Flasks
Brand:
Pyrex
YO-34554-23
More Details
Erlenmeyer flask mantle for 250-ml flask, 150 watts, 230 VAC (100AO917)
Category: Glas-Col Flexible Fabric Heating Mantles
Description: Erlenmeyer Flask Mantles
Brand:
Glas-Col
Model Number:
100AO917
YO-36227-25
More Details
Erlenmeyer flask mantle for 250-ml flask, 150 watts, 115 VAC (100AO916)
Category: Glas-Col Flexible Fabric Heating Mantles
Description: Erlenmeyer Flask Mantles
Brand:
Glas-Col
Model Number:
100AO916
YO-36227-28
More Details
Thermo Scientific Nalgene polypropylene Erlenmeyer flask, 500 mL
Category: Plastic Erlenmeyer Flasks
Description: Thermo Scientific Nalgene Polypropylene Erlenmeyer Flasks
Brand:
Thermo Scientific Nalgene
YO-06109-40
More Details
Thermo Scientific Nalgene polypropylene Erlenmeyer flask, 1000 mL
Category: Plastic Erlenmeyer Flasks
Description: Thermo Scientific Nalgene Polypropylene Erlenmeyer Flasks
Brand:
Thermo Scientific Nalgene
YO-06109-50
More Details
Thermo Scientific Nalgene polypropylene Erlenmeyer flask, 2000 mL
Category: Plastic Erlenmeyer Flasks
Description: Thermo Scientific Nalgene Polypropylene Erlenmeyer Flasks
Brand:
Thermo Scientific Nalgene
YO-06109-60
More Details
Chemware Erlenmeyer Flask, PFA, 100 mL
Category: Plastic Erlenmeyer Flasks
Description: Chemware Erlenmeyer Perfluoroalkoxy Flasks
Brand:
Chemware
YO-06112-00
More Details
Chemware Erlenmeyer Flask, PFA, 200 mL
Category: Plastic Erlenmeyer Flasks
Description: Chemware Erlenmeyer Perfluoroalkoxy Flasks
Brand:
Chemware
YO-06112-05
More Details
250 Ml Erlenmeyer Flask
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Kimax Glass
|
Next: 50 Ml Kjeldahl Flask
>
31125329-s-content_2 31125329scontent2