3 Methyl 1 Pentene

Image
  Product Detail
SKU

3-Methyl-1-pentene 99% (5g) (AC149980050)

Category: Methylp to Methylq
Description: 3-Methyl-1-pentene
Brand: Cole-Parmer


2-Methyl-1-pentene 99% (10ml) (AC127400100)

Category: Methylp to Methylq
Description: 2-Methyl-1-pentene
Brand: Cole-Parmer


2-Methyl-1-pentene 99% (50ml) (AC127400500)

Category: Methylp to Methylq
Description: 2-Methyl-1-pentene
Brand: Cole-Parmer


3-Methyl-1-butanol 98% (2.5l) (AC126480025)

Category: Methyl to Methylb
Description: 3-Methyl-1-butanol
Brand: Cole-Parmer


3-Methyl-1-butanol 98% (1l) (AC126480010)

Category: Methyl to Methylb
Description: 3-Methyl-1-butanol
Brand: Cole-Parmer


3-Methyl-1-butanol 98% (500ml) (AC126485000)

Category: Methyl to Methylb
Description: 3-Methyl-1-butanol
Brand: Cole-Parmer


3-Methyl-1-butanethiol 96% (25g) (AC152050250)

Category: Methyl to Methylb
Description: 3-Methyl-1-butanethiol
Brand: Cole-Parmer


3-Methyl-1-butanethiol 96% (100g) (AC152051000)

Category: Methyl to Methylb
Description: 3-Methyl-1-butanethiol
Brand: Cole-Parmer


4-Methyl-1-pentene 97% (25ml) (AC127420250)

Category: Methylp to Methylq
Description: 4-Methyl-1-pentene
Brand: Cole-Parmer


Poly(4-methyl-1-pentene), low molecular weight (500g) (AC184285000)

Category: Platina to Potash
Description: Poly(4-methyl-1-pentene)
Brand: Cole-Parmer


Poly(4-methyl-1-pentene), high molecular weight (500g) (AC184305000)

Category: Platina to Potash
Description: Poly(4-methyl-1-pentene)
Brand: Cole-Parmer


3-Methyl-1-phenyl-2-pyrazolin-5-one 99% (100g) (AC127461000)

Category: Methylp to Methylq
Description: 3-Methyl-1-phenyl-2-pyrazolin-5-one
Brand: Cole-Parmer


3-Methyl-1-phenyl-2-pyrazolin-5-one 99% (500g) (AC127465000)

Category: Methylp to Methylq
Description: 3-Methyl-1-phenyl-2-pyrazolin-5-one
Brand: Cole-Parmer


DL-2-Amino-3-methyl-1-butanol 95% (5g) (AC180420050)

Category: Aminom
Description: DL-2-Amino-3-methyl-1-butanol
Brand: Cole-Parmer


(S)-(+)-2-Amino-3-methyl-1-butanol 96% (1ml) (AC181590010)

Category: Aminom
Description: (S)-(+)-2-Amino-3-methyl-1-butanol
Brand: Cole-Parmer


5-Chloro-3-methyl-1-phenylpyrazole-4-carboxaldehyde 99% (1g) (AC333750010)

Category: Chlorom to Chloroo
Description: 5-Chloro-3-methyl-1-phenylpyrazole-4-carboxaldehyde
Brand: Cole-Parmer


5-Chloro-3-methyl-1-phenylpyrazole-4-carboxaldehyde 99% (5g) (AC333750050)

Category: Chlorom to Chloroo
Description: 5-Chloro-3-methyl-1-phenylpyrazole-4-carboxaldehyde
Brand: Cole-Parmer


3-Methyl-1-pentanol (5g) (AC335320050)

Category: Methylp to Methylq
Description: 3-Methyl-1-pentanol
Brand: Cole-Parmer


4-Carbethoxy-2-ethyl-3-methyl-2-cyclohexen-1-one tech. (25g) (AC155680250)

Category: Camphan to Carbowa
Description: 4-Carbethoxy-2-ethyl-3-methyl-2-cyclohexen-1-one
Brand: Cole-Parmer


Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride 97% (25g) (AC210560250)

Category: Methyl to Methylb
Description: Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride
Brand: Cole-Parmer




3 Methyl 1 Pentene
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended |  Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended

Previous: 3 Methyl 2 Butanone  |  Next: Cole-Parmer 3-Methyl-1-Butanol