4 Inch Brass Flange Fittings

Image
  Product Detail
SKU

Replacement Compression Ferrule and Nut Assembly, for 1/4" OD, Brass

Category: Compression Nut Assemblies
Description: Brass compression nut assemblies (with ferrules)
Brand: Cole-Parmer


Barbed Elbow, 1/4 Inch, Antimicrobial Polypropylene, Pack Of 5 (L0-4 AGNP)

Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Brand: Cole-Parmer
Model Number: L0-4 AGNP


Barbed Tee, 1/4 Inch, Antimicrobial Polypropylene, Pack Of 5 (T0-4 AGNP)

Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Model Number: T0-4 AGNP


Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x 1/4-inch tubing ID

Category: Cole-Parmer 316L Stainless Steel Sanitary Clamp Couplings
Description: Cole-Parmer 316L Stainless Steel Barbed Sanitary Clamp Couplings
Brand: Cole-Parmer


Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x 3/4-inch tubing ID

Category: Cole-Parmer 316L Stainless Steel Sanitary Clamp Couplings
Description: Cole-Parmer 316L Stainless Steel Barbed Sanitary Clamp Couplings
Brand: Cole-Parmer


Compression male pipe adapter elbow, 1/4" NPT(M) x 1/4" OD, brass

Category: Male Threaded Adapter Elbows
Description: Brass Compression to NPT(M) Adapter Elbows
Brand: Cole-Parmer


Compression male pipe adapter elbow, 3/4" NPT(M) x 3/4" OD, brass

Category: Male Threaded Adapter Elbows
Description: Brass Compression to NPT(M) Adapter Elbows
Brand: Cole-Parmer


Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x 1/8-inch tubing ID

Category: Cole-Parmer 316L Stainless Steel Sanitary Clamp Couplings
Description: Cole-Parmer 316L Stainless Steel Barbed Sanitary Clamp Couplings
Brand: Cole-Parmer


Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x 3/8-inch tubing ID

Category: Cole-Parmer 316L Stainless Steel Sanitary Clamp Couplings
Description: Cole-Parmer 316L Stainless Steel Barbed Sanitary Clamp Couplings
Brand: Cole-Parmer


Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x 1/2-inch tubing ID

Category: Cole-Parmer 316L Stainless Steel Sanitary Clamp Couplings
Description: Cole-Parmer 316L Stainless Steel Barbed Sanitary Clamp Couplings
Brand: Cole-Parmer


Compression male pipe adapter elbow, 1/8" NPT(M) x 1/4" OD, brass

Category: Male Threaded Adapter Elbows
Description: Brass Compression to NPT(M) Adapter Elbows
Brand: Cole-Parmer


Compression male pipe adapter elbow, 3/8" NPT(M) x 1/4" OD, brass

Category: Male Threaded Adapter Elbows
Description: Brass Compression to NPT(M) Adapter Elbows
Brand: Cole-Parmer


Barbed Elbow, 3/4 Inch, Antimicrobial Polypropylene, Pack Of 5 (L0-12 AGNP)

Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Model Number: L0-12 AGNP


Barbed Tee, 3/4 Inch, Antimicrobial Polypropylene, Pack Of 5 (T0-12 AGNP)

Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Model Number: T0-12 AGNP


BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06368-73)

Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Brand: Cole-Parmer


BARBED ELBOW, 3/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06368-76)

Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Brand: Cole-Parmer


BARBED TEE, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06368-82)

Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Brand: Cole-Parmer


BARBED TEE, 3/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06368-85)

Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Brand: Cole-Parmer


Barbed Fittings; Cross Connector, Clear PP, 1/4-inch Tubing ID, 50/pk (YO-31417-11)

Category: Barbed to barbed connectors
Description: Barbed cross connectors
Brand: Cole-Parmer


Barbed Y-Connectors, POLYPROPYLENE, 3/4-INCH, PACK OF 5

Category: Barbed to barbed connectors
Description: Barbed Y connectors
Brand: Cole-Parmer
Model Number: Y0-12NP (5 PACK)




4 Inch Brass Flange Fittings
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended |  Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended

Previous: Fitting M3 to 1 4 Inch  |  Next: 4 Inch Plastic Bulkhead Fittings