Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Related Categories
Chest Freezers
Revco Chest Freezer
Scientemp Freezers - Chest/Cabinet
Thermo Chest Freezers
Freezers - Chest/Cabinet
Chest Freezer Temperature Controller
Cole-Parmer Action Back Coveralls
Revco Freezer Rel 2304 V 20
2.0 Cubic Feet Lab Incubator
Cole Po
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
40 Chest
Image
Product Detail
SKU
Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-D)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT1350-10-D
YO-44147-04
More Details
Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-V)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT1350-10-V
YO-44147-05
More Details
Thermo Scientific Revco CxF -40°C Chest Freezer, 17 cubic feet, 11
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT1750-10-A
YO-44147-06
More Details
Thermo Scientific Revco CxF -40°C Chest Freezer, 17 cubic feet, 20
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT1750-10-D
YO-44147-07
More Details
Thermo Scientific Revco CxF -40°C Chest Freezer, 17 cubic feet, 23
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT1750-10-V
YO-44147-08
More Details
Thermo Scientific Revco CxF -40°C Chest Freezer, 20 cubic feet, 11
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT2050-10-A
YO-44147-09
More Details
Thermo Scientific Revco CxF -40°C Chest Freezer, 20 cubic feet, 20
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT2050-10-D
YO-44147-10
More Details
Thermo Scientific Revco CxF -40°C Chest Freezer, 20 cubic feet, 23
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT2050-10-V
YO-44147-11
More Details
Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-A)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT1350-10-A
YO-44147-03
More Details
Thermo Scientific Revco CxF -40°C Chest Freezer, 3 cubic feet (3 c
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT350-10-V
YO-44147-02
More Details
Thermo Scientific Revco CxF -40°C Series, Chest Freezer, 3 cubic
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers
Brand:
Thermo Scientific Revco
Model Number:
ULT350-10-A
YO-44147-01
More Details
Standard low-temperature cabinets; 9.4 cu ft; -40°C; 230V (40-9.4 230V/50HZ)
Category: Chest-style Freezers
Description: ScienTemp Standard Low-Temperature Cabinets
Brand:
ScienTemp
Model Number:
40-9.4 230V/50HZ
YO-03815-12
More Details
Standard low-temperature cabinets; 9.4 cu ft; -40°C; 115V (40-9.4)
Category: Chest-style Freezers
Description: ScienTemp Standard Low-Temperature Cabinets
Brand:
ScienTemp
Model Number:
40-9.4
YO-03815-22
More Details
Action back coveralls; Size, medium; chest size, 38" to 40"; color, na
Category: Coveralls
Description: Action Back Coveralls
Brand:
Cole-Parmer
Model Number:
CT10NV-MEDIUM,(38-40)
YO-86296-10
More Details
Action back coveralls; size, medium; chest size, 38" to 40"; color, po
Category: Coveralls
Description: Action Back Coveralls
Brand:
Cole-Parmer
Model Number:
CT10PB-MEDIUM,(38-40)
YO-86296-20
More Details
Action back coveralls; size, medium; chest size, 38" to 40"; color, ch
Category: Coveralls
Description: Action Back Coveralls
Brand:
Cole-Parmer
Model Number:
CT10CH-MEDIUM,(38-40)
YO-86296-30
More Details
40 Chest
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Revco Freezer Ult1390
|
Next: 2.0 Cubic Feet Lab Incubator
>
31125329-s-content_2 31125329scontent2