5 Gallon Chemical Containers

Image
  Product Detail
SKU

3M Folded Sorbents, Chemical kit (C-SKFL5)

Category: Spill Cleanup
Description: 3M Folded Sorbents
Brand: 3M


Tetrabutylammonium chloride (contains alsobromide) tech., 95% (5g) (AC174120050)

Category: Tertraa to Tetrae
Description: Tetrabutylammonium chloride (contains alsobromide)
Brand: Cole-Parmer


Tetrabutylammonium chloride (contains alsobromide) tech., 95% (25g) (AC174120250)

Category: Tertraa to Tetrae
Description: Tetrabutylammonium chloride (contains alsobromide)
Brand: Cole-Parmer


Tetrabutylammonium chloride (contains alsobromide) tech., 95% (100g) (AC174121000)

Category: Tertraa to Tetrae
Description: Tetrabutylammonium chloride (contains alsobromide)
Brand: Cole-Parmer


4-(4-Fluorobenzoyl)piperidine p-toluenesulfonate, contains HCl-salt 95% (5g)

Category: Fluoro to Fluoron
Description: 4-(4-Fluorobenzoyl)piperidine p-toluenesulfonate, contains HCl-salt
Brand: Cole-Parmer


Tetrahexylammonium benzoate (contains various amounts of water) (25g) (AC420390250)

Category: Tetraf to Tetral
Description: Tetrahexylammonium benzoate (contains various amounts of water)
Brand: Cole-Parmer


Thermo Scientific Nalgene Biohazard Waste Container, 5 gal.

Category: Spill Cleanup
Description: Thermo Scientific Nalgene Biohazard Waste Containers
Brand: Thermo Scientific Nalgene


Chemical Splash Suit, Medium

Category: Coveralls
Description: Chemical Splash Suits
Brand: Cole-Parmer


Chemical Splash Suit, Large

Category: Coveralls
Description: Chemical Splash Suits
Brand: Cole-Parmer


Chemical Splash Suit, X-Large

Category: Coveralls
Description: Chemical Splash Suits
Brand: Cole-Parmer


Container, HDPE Box without Lid; 5 gallon

Category: Boxes, Totes, and Lids
Description: Boxes, HDPE
Brand: Cole-Parmer


Shipping Carton for Collapsible Container 06100-30

Category: Plastic Containers
Description: Shipping Cartons for Collapsible Containers
Brand: Guaranteed In Stock Program


Tubing, Masterflex, Tygon Chemical, L/S 13, 50 ft

Category: L/S Precision Pump Tubing for Masterflex L/S Pumps
Description: L/S Precision Tygon Chemical (2001) Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Tubing, Masterflex, Tygon Chemical, L/S 14, 50 ft

Category: L/S Precision Pump Tubing for Masterflex L/S Pumps
Description: L/S Precision Tygon Chemical (2001) Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Tubing, Masterflex, Tygon Chemical, L/S15, 50 ft

Category: L/S High-Performance Precision Pump Tubing for Masterflex L/S Pumps
Description: L/S High-Performance Precision Tygon Chemical (2001) Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Tubing, Masterflex, Tygon Chemical, L/S 16, 50 ft

Category: L/S Precision Pump Tubing for Masterflex L/S Pumps
Description: L/S Precision Tygon Chemical (2001) Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Tubing, Masterflex, Tygon Chemical, L/S 17, 50 ft

Category: L/S Precision Pump Tubing for Masterflex L/S Pumps
Description: L/S Precision Tygon Chemical (2001) Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Tubing, Masterflex, Tygon Chemical, L/S 18, 50 ft

Category: L/S Precision Pump Tubing for Masterflex L/S Pumps
Description: L/S Precision Tygon Chemical (2001) Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Tubing, Masterflex, Tygon Chemical, L/S24, 50 ft

Category: L/S High-Performance Precision Pump Tubing for Masterflex L/S Pumps
Description: L/S High-Performance Precision Tygon Chemical (2001) Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Tubing, Masterflex, Tygon Chemical, L/S 25, 50 ft

Category: L/S Precision Pump Tubing for Masterflex L/S Pumps
Description: L/S Precision Tygon Chemical (2001) Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex




5 Gallon Chemical Containers
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended |  Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended

Previous: Kynar Sanitary L/S Tubing  |  Next: Chemical Compatibility Chart